Learn More
4-Benzyloxyaniline hydrochloride, 98%, Thermo Scientific Chemicals
$167.68 - $167.68
Chemical Identifiers
CAS | 51388-20-6 |
---|---|
Molecular Formula | C13H13NO·HCl |
Molecular Weight (g/mol) | 235.72 |
MDL Number | MFCD00012995 |
InChI Key | KQBDLOVXZHOAJI-UHFFFAOYSA-N |
Synonym | 4-benzyloxyaniline hydrochloride, 4-benzyloxy aniline hydrochloride, 4-benzyloxyaniline hcl, benzenamine, 4-phenylmethoxy-, hydrochloride, 4-phenylmethoxyaniline hydrochloride, aniline, p-benzyloxy-hydrochloride, 4-benzyloxy phenyl amine hydrochloride, 4-phenylmethoxy phenylamine, chloride, acmc-20amop, pubchem21343 |
PubChem CID | 2723831 |
IUPAC Name | 4-phenylmethoxyaniline;hydrochloride |
SMILES | C1=CC=C(C=C1)COC2=CC=C(C=C2)N.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC152291000
|
Thermo Scientific Chemicals
152291000 |
100 g | Plastic bottle |
Each for $167.68
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
51388-20-6 | |
235.72 | |
KQBDLOVXZHOAJI-UHFFFAOYSA-N | |
2723831 | |
C1=CC=C(C=C1)COC2=CC=C(C=C2)N.Cl |
C13H13NO·HCl | |
MFCD00012995 | |
4-benzyloxyaniline hydrochloride, 4-benzyloxy aniline hydrochloride, 4-benzyloxyaniline hcl, benzenamine, 4-phenylmethoxy-, hydrochloride, 4-phenylmethoxyaniline hydrochloride, aniline, p-benzyloxy-hydrochloride, 4-benzyloxy phenyl amine hydrochloride, 4-phenylmethoxy phenylamine, chloride, acmc-20amop, pubchem21343 | |
4-phenylmethoxyaniline;hydrochloride |
Specifications
51388-20-6 | |
100.0 | |
Brown | |
98% | |
C13H13NO·HCl | |
MFCD00012995 | |
13, 439 | |
Solubility in water: slightly soluble | |
C1=CC=C(C=C1)COC2=CC=C(C=C2)N.Cl | |
235.72 | |
235.72 | |
Crystalline Powder |
97.5 | |
228°C | |
Authentic | |
Plastic bottle | |
C6H5CH2OC6H4NH2·HCl | |
100 g | |
4-benzyloxyaniline hydrochloride, 4-benzyloxy aniline hydrochloride, 4-benzyloxyaniline hcl, benzenamine, 4-phenylmethoxy-, hydrochloride, 4-phenylmethoxyaniline hydrochloride, aniline, p-benzyloxy-hydrochloride, 4-benzyloxy phenyl amine hydrochloride, 4-phenylmethoxy phenylamine, chloride, acmc-20amop, pubchem21343 | |
KQBDLOVXZHOAJI-UHFFFAOYSA-N | |
4-phenylmethoxyaniline;hydrochloride | |
2723831 | |
98% | |
4-Benzyloxyaniline hydrochloride, 98% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 257-170-6
RUO – Research Use Only