missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Benzyloxybenzotrifluoride, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H61303MD

 View more versions of this product

Catalog No. AAH61303MD



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-Benzyloxybenzotrifluoride
C14H11F3O
250 mg
SNRFAICCYVUTRG-UHFFFAOYSA-N
1-phenylmethoxy-4-(trifluoromethyl)benzene
2798168
95%
70097-65-3
MFCD00275905
1-benzyloxy-4-trifluoromethyl benzene, 4-benzyloxybenzotrifluoride, 4-benzyloxy benzotrifluoride, 1-phenylmethoxy-4-trifluoromethyl benzene, maybridge1_001022, 4 benzyloxy benzotrifluoride, 4-trifluoromethylphenyl benzyl ether, 1-benzyloxy-4-trifluoromethylbenzene, 1-benzyloxy-4-trifluoromethyl-benzene
C1=CC=C(C=C1)COC2=CC=C(C=C2)C(F)(F)F
252.236
252.23
Solid

Chemical Identifiers

70097-65-3
252.236
SNRFAICCYVUTRG-UHFFFAOYSA-N
2798168
C1=CC=C(C=C1)COC2=CC=C(C=C2)C(F)(F)F
C14H11F3O
MFCD00275905
1-benzyloxy-4-trifluoromethyl benzene, 4-benzyloxybenzotrifluoride, 4-benzyloxy benzotrifluoride, 1-phenylmethoxy-4-trifluoromethyl benzene, maybridge1_001022, 4 benzyloxy benzotrifluoride, 4-trifluoromethylphenyl benzyl ether, 1-benzyloxy-4-trifluoromethylbenzene, 1-benzyloxy-4-trifluoromethyl-benzene
1-phenylmethoxy-4-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions