missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Benzyloxyphenoxyacetic acid, 95%, Thermo Scientific™

$496.62 - $496.62

Chemical Identifiers

CAS 38559-92-1
Molecular Formula C15H14O4
Molecular Weight (g/mol) 258.273
MDL Number MFCD00014361
InChI Key VXMSXBVTUNOSLL-UHFFFAOYSA-N
Synonym 4-benzyloxyphenoxyacetic acid, 4-benzyloxy phenoxy acetic acid, 4-benzyloxy phenoxyacetic acid, 2-4-benzyloxy phenoxy acetic acid, 4-phenylmethoxy phenoxy acetic acid, acetic acid, 4-phenylmethoxy phenoxy, 2-4-phenylmethoxy phenoxy acetic acid, enamine_002147, acmc-1agp8
PubChem CID 563696
IUPAC Name 2-(4-phenylmethoxyphenoxy)acetic acid
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)OCC(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0104718
View Documents
Thermo Scientific Chemicals
L0104718
50 g
Each for $496.62
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

38559-92-1
258.273
VXMSXBVTUNOSLL-UHFFFAOYSA-N
563696
C1=CC=C(C=C1)COC2=CC=C(C=C2)OCC(=O)O
C15H14O4
MFCD00014361
4-benzyloxyphenoxyacetic acid, 4-benzyloxy phenoxy acetic acid, 4-benzyloxy phenoxyacetic acid, 2-4-benzyloxy phenoxy acetic acid, 4-phenylmethoxy phenoxy acetic acid, acetic acid, 4-phenylmethoxy phenoxy, 2-4-phenylmethoxy phenoxy acetic acid, enamine_002147, acmc-1agp8
2-(4-phenylmethoxyphenoxy)acetic acid

Specifications

38559-92-1
C15H14O4
50 g
4-benzyloxyphenoxyacetic acid, 4-benzyloxy phenoxy acetic acid, 4-benzyloxy phenoxyacetic acid, 2-4-benzyloxy phenoxy acetic acid, 4-phenylmethoxy phenoxy acetic acid, acetic acid, 4-phenylmethoxy phenoxy, 2-4-phenylmethoxy phenoxy acetic acid, enamine_002147, acmc-1agp8
C1=CC=C(C=C1)COC2=CC=C(C=C2)OCC(=O)O
258.273
258.28
4-Benzyloxyphenoxyacetic acid
139°C to 140°C
MFCD00014361
2469260
VXMSXBVTUNOSLL-UHFFFAOYSA-N
2-(4-phenylmethoxyphenoxy)acetic acid
563696
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 254-000-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'