missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Boc-amino)-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H5919403

 View more versions of this product

Catalog No. AAH5919403


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is the key intermediate in the synthesis of ,3-Dihydro-1,4-benzodiazepin-2(2H)-ones and 3-Amino-1,3-dihydro-1,4-benzodiazepin-2(2H)-ones. This synthesis is done with Pd-catalyzed cross-coupling of imidoyl chlorides with organoboronic acids.

Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications

Specifications

4-(Boc-amino)-2-fluorobenzeneboronic acid pinacol ester
150°C to 152°C
MFCD18383441
tert-butyl 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl n-3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 4-boc-amino-2-fluorobenzeneboronic acid pinacol ester, 3-fluoro-4-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl phenyl carbamic acid tert-butyl ester
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)NC(=O)OC(C)(C)C)F
337.198
337.2
1256256-45-7
C17H25BFNO4
1 g
FALNYBWTRBHWID-UHFFFAOYSA-N
tert-butyl N-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
59578631
96%

Chemical Identifiers

1256256-45-7
337.198
FALNYBWTRBHWID-UHFFFAOYSA-N
59578631
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)NC(=O)OC(C)(C)C)F
C17H25BFNO4
MFCD18383441
tert-butyl 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl n-3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 4-boc-amino-2-fluorobenzeneboronic acid pinacol ester, 3-fluoro-4-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl phenyl carbamic acid tert-butyl ester
tert-butyl N-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions