missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Boc-aminomethyl)-3-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
$624.92 - $624.92
Chemical Identifiers
| CAS | 1351501-44-4 |
|---|---|
| Molecular Formula | C18H27BFNO4 |
| Molecular Weight (g/mol) | 351.225 |
| InChI Key | SAUVAJYHMCIQNE-UHFFFAOYSA-N |
| Synonym | 4-(Boc-aminomethyl)-3-fluorophenylboronic acid pinacol ester |
| PubChem CID | 66980292 |
| IUPAC Name | tert-butyl N-[[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]carbamate |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)CNC(=O)OC(C)(C)C)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6294803
|
Thermo Scientific Chemicals
H6294803 |
1 g |
Each for $624.92
|
|
|||||
Description
4-(Boc-aminomethyl)-3-fluorobenzeneboronic acid generally used in the synthesis of organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Boc-aminomethyl)-3-fluorobenzeneboronic acid generally used in the synthesis of organic compounds.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
| 1351501-44-4 | |
| 351.225 | |
| 4-(Boc-aminomethyl)-3-fluorophenylboronic acid pinacol ester | |
| tert-butyl N-[[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]carbamate |
| C18H27BFNO4 | |
| SAUVAJYHMCIQNE-UHFFFAOYSA-N | |
| 66980292 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)CNC(=O)OC(C)(C)C)F |
Specifications
| 1351501-44-4 | |
| 1 g | |
| SAUVAJYHMCIQNE-UHFFFAOYSA-N | |
| tert-butyl N-[[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]carbamate | |
| 66980292 | |
| 96% |
| C18H27BFNO4 | |
| 4-(Boc-aminomethyl)-3-fluorophenylboronic acid pinacol ester | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)CNC(=O)OC(C)(C)C)F | |
| 351.225 | |
| 351.22 | |
| 4-(Boc-aminomethyl)-3-fluorobenzeneboronic acid pinacol ester |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only