missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromo-1-methyl-1H-pyrazol-3-yl)methanol, ≥97%, Thermo Scientific™

$360.54 - $360.54

Chemical Identifiers

CAS 915707-65-2
Molecular Formula C5H7BrN2O
Molecular Weight (g/mol) 191.028
MDL Number MFCD06202888
InChI Key JFGLJTFTVBZOCB-UHFFFAOYSA-N
Synonym 4-bromo-1-methyl-1h-pyrazol-3-yl methanol, 4-bromo-1-methylpyrazol-3-yl methanol, 4-bromo-3-hydroxymethyl-1-methyl-1h-pyrazole, 4-bromo-1-methylpyrazol-3-yl methan-1-ol, pubchem23717
PubChem CID 24229713
IUPAC Name (4-bromo-1-methylpyrazol-3-yl)methanol
SMILES CN1C=C(C(=N1)CO)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC63909DA
SDS View Documents
Thermo Scientific Chemicals
CC63909DA
1 g
Each for $360.54
Only null left
 
Description

Description

Specifications

Chemical Identifiers

915707-65-2
191.028
JFGLJTFTVBZOCB-UHFFFAOYSA-N
24229713
CN1C=C(C(=N1)CO)Br
C5H7BrN2O
MFCD06202888
4-bromo-1-methyl-1h-pyrazol-3-yl methanol, 4-bromo-1-methylpyrazol-3-yl methanol, 4-bromo-3-hydroxymethyl-1-methyl-1h-pyrazole, 4-bromo-1-methylpyrazol-3-yl methan-1-ol, pubchem23717
(4-bromo-1-methylpyrazol-3-yl)methanol

Specifications

915707-65-2
100.0
Amber Glass Bottle
MFCD06202888
4-bromo-1-methyl-1h-pyrazol-3-yl methanol, 4-bromo-1-methylpyrazol-3-yl methanol, 4-bromo-3-hydroxymethyl-1-methyl-1h-pyrazole, 4-bromo-1-methylpyrazol-3-yl methan-1-ol, pubchem23717
CN1C=C(C(=N1)CO)Br
191.028
191.03
(4-Bromo-1-methyl-1H-pyrazol-3-yl)methanol
96.5
97+%
C5H7BrN2O
1 g
JFGLJTFTVBZOCB-UHFFFAOYSA-N
(4-bromo-1-methylpyrazol-3-yl)methanol
24229713
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents