Learn More
4-Bromo-2,2-diphenylbutyronitrile, 95%
CAS: 39186-58-8 | C16H14BrN | 300.199 g/mol
$231.26 - $990.34
Chemical Identifiers
| CAS | 39186-58-8 |
|---|---|
| Molecular Formula | C16H14BrN |
| Molecular Weight (g/mol) | 300.199 |
| MDL Number | MFCD00001845 |
| InChI Key | IGYSFJHVFHNOEI-UHFFFAOYSA-N |
| Synonym | 4-bromo-2,2-diphenylbutyronitrile, 2,2-diphenyl-4-bromobutyronitrile, 4-bromo-2,2-diphenyl-butyronitrile, 3-cyano-3,3-diphenylpropyl bromide, benzeneacetonitrile, .alpha.-2-bromoethyl-.alpha.-phenyl, acmc-1csza, butyronitrile,2-diphenyl, ksc495i2r, aronis25014, bromoethyl diphenyl acetonitrile |
| PubChem CID | 96575 |
| IUPAC Name | 4-bromo-2,2-diphenylbutanenitrile |
| SMILES | C1=CC=C(C=C1)C(CCBr)(C#N)C2=CC=CC=C2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1778622
|
Thermo Scientific Chemicals
A1778622 |
100 g |
Each for $231.26
|
|
|||||
|
AAA1778636
|
Thermo Scientific Chemicals
A1778636 |
500 g |
Each for $990.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 39186-58-8 | |
| 300.199 | |
| IGYSFJHVFHNOEI-UHFFFAOYSA-N | |
| 96575 | |
| C1=CC=C(C=C1)C(CCBr)(C#N)C2=CC=CC=C2 |
| C16H14BrN | |
| MFCD00001845 | |
| 4-bromo-2,2-diphenylbutyronitrile, 2,2-diphenyl-4-bromobutyronitrile, 4-bromo-2,2-diphenyl-butyronitrile, 3-cyano-3,3-diphenylpropyl bromide, benzeneacetonitrile, .alpha.-2-bromoethyl-.alpha.-phenyl, acmc-1csza, butyronitrile,2-diphenyl, ksc495i2r, aronis25014, bromoethyl diphenyl acetonitrile | |
| 4-bromo-2,2-diphenylbutanenitrile |
Specifications
| 39186-58-8 | |
| C16H14BrN | |
| 100 g | |
| 1684952 | |
| IGYSFJHVFHNOEI-UHFFFAOYSA-N | |
| 4-bromo-2,2-diphenylbutanenitrile | |
| 96575 | |
| 95% |
| 67°C to 69°C | |
| MFCD00001845 | |
| UN3439 | |
| 4-bromo-2,2-diphenylbutyronitrile, 2,2-diphenyl-4-bromobutyronitrile, 4-bromo-2,2-diphenyl-butyronitrile, 3-cyano-3,3-diphenylpropyl bromide, benzeneacetonitrile, .alpha.-2-bromoethyl-.alpha.-phenyl, acmc-1csza, butyronitrile,2-diphenyl, ksc495i2r, aronis25014, bromoethyl diphenyl acetonitrile | |
| C1=CC=C(C=C1)C(CCBr)(C#N)C2=CC=CC=C2 | |
| 300.199 | |
| 300.2 | |
| 4-Bromo-2,2-diphenylbutyronitrile |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P301+P312-P330-P501c
H302
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 254-337-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only