missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2,3-difluorophenol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2636906

 View more versions of this product

Catalog No. AAH2636906


Explore available promotions

Description

Description

4-Bromo-2,3-difluorophenol is used as medical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2,3-difluorophenol is used as medical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

4-Bromo-2,3-difluorophenol
C6H3BrF2O
5 g
JZAVCMMYGSROJP-UHFFFAOYSA-N
4-bromo-2,3-difluorophenol
10219942
97%
144292-32-0
MFCD08061907
2,3-difluoro-4-bromophenol, 4-bromo-2,3-difluoro-phenol, phenol, 4-bromo-2,3-difluoro, pubchem2477, pubchem4111, acmc-209csf, bromodifluorophenol, ksc494o1n, 2,3-difluoro 4-bromo phenol, 2.3-difluoro-4-bromo phenol
OC1=C(F)C(F)=C(Br)C=C1
208.99
208.99

Chemical Identifiers

144292-32-0
208.99
JZAVCMMYGSROJP-UHFFFAOYSA-N
10219942
OC1=C(F)C(F)=C(Br)C=C1
C6H3BrF2O
MFCD08061907
2,3-difluoro-4-bromophenol, 4-bromo-2,3-difluoro-phenol, phenol, 4-bromo-2,3-difluoro, pubchem2477, pubchem4111, acmc-209csf, bromodifluorophenol, ksc494o1n, 2,3-difluoro 4-bromo phenol, 2.3-difluoro-4-bromo phenol
4-bromo-2,3-difluorophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only