missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2,5-difluorophenol, 99%, Thermo Scientific Chemicals

$168.62 - $487.12

Chemical Identifiers

CAS 486424-36-6
Molecular Formula C6H3BrF2O
Molecular Weight (g/mol) 208.99
MDL Number MFCD04973751
InChI Key BYZMZJIWCQTYSR-UHFFFAOYSA-N
Synonym 1-bromo-2,5-difluoro-4-hydroxybenzene, 4-bromo-2,5-difluoro-phenol, phenol, 4-bromo-2,5-difluoro, acmc-20amnw, phenol,4-bromo-2,5-difluoro, 4-bromanyl-2,5-bis fluoranyl phenol, 1-hydroxy-4-bromo-2,5-difluoro-benzene
PubChem CID 7018042
IUPAC Name 4-bromo-2,5-difluorophenol
SMILES C1=C(C(=CC(=C1F)Br)F)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2621206
View Documents
Thermo Scientific Chemicals
H2621206
5 g
Each for $168.62
 
AAH2621214
View Documents
Thermo Scientific Chemicals
H2621214
25 g
Each for $487.12
 
Description

Description

4-Bromo-2,5-difluorophenol is used as a raw material in organic synthesis. Used in the synthesis allyloxy-based biphenyl liquid crystals with multiple lateral fluoro substituents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2,5-difluorophenol is used as a raw material in organic synthesis. Used in the synthesis allyloxy-based biphenyl liquid crystals with multiple lateral fluoro substituents.

Solubility
Soluble in water (slightly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

486424-36-6
208.99
BYZMZJIWCQTYSR-UHFFFAOYSA-N
7018042
C1=C(C(=CC(=C1F)Br)F)O
C6H3BrF2O
MFCD04973751
1-bromo-2,5-difluoro-4-hydroxybenzene, 4-bromo-2,5-difluoro-phenol, phenol, 4-bromo-2,5-difluoro, acmc-20amnw, phenol,4-bromo-2,5-difluoro, 4-bromanyl-2,5-bis fluoranyl phenol, 1-hydroxy-4-bromo-2,5-difluoro-benzene
4-bromo-2,5-difluorophenol

Specifications

486424-36-6
C6H3BrF2O
5 g
Soluble in water (slightly).
C1=C(C(=CC(=C1F)Br)F)O
208.99
208.99
4-Bromo-2,5-difluorophenol
51°C to 53°C
MFCD04973751
1-bromo-2,5-difluoro-4-hydroxybenzene, 4-bromo-2,5-difluoro-phenol, phenol, 4-bromo-2,5-difluoro, acmc-20amnw, phenol,4-bromo-2,5-difluoro, 4-bromanyl-2,5-bis fluoranyl phenol, 1-hydroxy-4-bromo-2,5-difluoro-benzene
BYZMZJIWCQTYSR-UHFFFAOYSA-N
4-bromo-2,5-difluorophenol
7018042
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only