missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,6-dimethylanisole, 99%

CAS: 14804-38-7 | C9H11BrO | 215.09 g/mol

$26.12 - $87.38

Chemical Identifiers

CAS 14804-38-7
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD00143259
InChI Key MMARFGDTMJBIBK-UHFFFAOYSA-N
Synonym 4-bromo-2,6-dimethylanisole, 5-bromo-2-methoxy-m-xylene, 4-brom-2,6-dimethylanisole, 1-bromo-3,5-dimethyl-4-methoxybenzene, 2,6-dimethyl-4-bromoanisole, benzene, 5-bromo-2-methoxy-1,3-dimethyl, timtec-bb sbb005776, attercop-chm at108629, 4-bromo-2,6-dimethyl-methoxybenzene, pubchem2654
PubChem CID 278654
IUPAC Name 5-bromo-2-methoxy-1,3-dimethylbenzene
SMILES COC1=C(C)C=C(Br)C=C1C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2193106
View Documents
Thermo Scientific Chemicals
B2193106
5 g
Each for $26.12
Only null left
 
AAB2193114
View Documents
Thermo Scientific Chemicals
B2193114
25 g
Each for $87.38
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14804-38-7
215.09
MMARFGDTMJBIBK-UHFFFAOYSA-N
278654
COC1=C(C)C=C(Br)C=C1C
C9H11BrO
MFCD00143259
4-bromo-2,6-dimethylanisole, 5-bromo-2-methoxy-m-xylene, 4-brom-2,6-dimethylanisole, 1-bromo-3,5-dimethyl-4-methoxybenzene, 2,6-dimethyl-4-bromoanisole, benzene, 5-bromo-2-methoxy-1,3-dimethyl, timtec-bb sbb005776, attercop-chm at108629, 4-bromo-2,6-dimethyl-methoxybenzene, pubchem2654
5-bromo-2-methoxy-1,3-dimethylbenzene

Specifications

14804-38-7
117°C to 120°C (13 mmHg)
1.545
MFCD00143259
4-bromo-2,6-dimethylanisole, 5-bromo-2-methoxy-m-xylene, 4-brom-2,6-dimethylanisole, 1-bromo-3,5-dimethyl-4-methoxybenzene, 2,6-dimethyl-4-bromoanisole, benzene, 5-bromo-2-methoxy-1,3-dimethyl, timtec-bb sbb005776, attercop-chm at108629, 4-bromo-2,6-dimethyl-methoxybenzene, pubchem2654
COC1=C(C)C=C(Br)C=C1C
215.09
215.09
4-Bromo-2,6-dimethylanisole
1.347
5 g
C9H11BrO
1940290
MMARFGDTMJBIBK-UHFFFAOYSA-N
5-bromo-2-methoxy-1,3-dimethylbenzene
278654
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only