missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,6-dimethylphenol, 99%

CAS: 2374-05-2 | C8H9BrO | 201.06 g/mol

$68.68 - $639.04

Chemical Identifiers

CAS 2374-05-2
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD00002314
InChI Key ZLVFYUORUHNMBO-UHFFFAOYSA-N
Synonym 4-bromo-2,6-xylenol, phenol, 4-bromo-2,6-dimethyl, 2,6-dimethyl-4-bromophenol, 2,6-xylenol, 4-bromo, pubchem9579, 2, 4-bromo, acmc-2097aq, 4-bromo-2,6 dimethylphenol, 5-bromo-2-hydroxy-m-xylene, ksc495g0d
PubChem CID 16919
IUPAC Name 4-bromo-2,6-dimethylphenol
SMILES CC1=CC(=CC(=C1O)C)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2166914
View Documents
Thermo Scientific Chemicals
B2166914
25 g N/A
Only null left
 
AAB2166922
View Documents
Thermo Scientific Chemicals
B2166922
100 g N/A
Only null left
 
AAB2166936
View Documents
Thermo Scientific Chemicals
B2166936
500 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
It is used as a preservatives for coatings, slurries and to control microbial fouling in paper mills , oil field and leather process and water treatment process against microbial attack.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

2374-05-2
201.06
ZLVFYUORUHNMBO-UHFFFAOYSA-N
16919
CC1=CC(=CC(=C1O)C)Br
C8H9BrO
MFCD00002314
4-bromo-2,6-xylenol, phenol, 4-bromo-2,6-dimethyl, 2,6-dimethyl-4-bromophenol, 2,6-xylenol, 4-bromo, pubchem9579, 2, 4-bromo, acmc-2097aq, 4-bromo-2,6 dimethylphenol, 5-bromo-2-hydroxy-m-xylene, ksc495g0d
4-bromo-2,6-dimethylphenol

Specifications

2374-05-2
Phenol-like
MFCD00002314
1862399
Slightly soluble in water.
CC1=CC(=CC(=C1O)C)Br
201.06
201.07
4-Bromo-2,6-dimethylphenol
74°C to 78°C
C8H9BrO
25 g
4-bromo-2,6-xylenol, phenol, 4-bromo-2,6-dimethyl, 2,6-dimethyl-4-bromophenol, 2,6-xylenol, 4-bromo, pubchem9579, 2, 4-bromo, acmc-2097aq, 4-bromo-2,6 dimethylphenol, 5-bromo-2-hydroxy-m-xylene, ksc495g0d
ZLVFYUORUHNMBO-UHFFFAOYSA-N
4-bromo-2,6-dimethylphenol
16919
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 219-153-1

RTECSNumber : ZE6780000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only