missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-cyanopyridine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3442403

 View more versions of this product

Catalog No. AAH3442403


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromo-2-cyanopyridine
87°C to 91°C
MFCD04065805
UN3439
CZXDCTUSFIKLIJ-UHFFFAOYSA-N
4-bromopyridine-2-carbonitrile
693283
97%
62150-45-2
C6H3BrN2
1 g
4-bromo-2-cyanopyridine, 4-bromopicolinonitrile, 4-bromo-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 4-bromo, 2-cyano-4-bromopyridine, 4-bromo-2-pyridinecarbonitrile, pubchem23829, 4-bromo-2-cyano-pyridine, 2-pyridinecarbonitrile,4-bromo, abbypharma ap-17-5586
C1=CN=C(C=C1Br)C#N
183.008
183.01

Chemical Identifiers

62150-45-2
183.008
CZXDCTUSFIKLIJ-UHFFFAOYSA-N
4-bromopyridine-2-carbonitrile
C6H3BrN2
MFCD04065805
693283
C1=CN=C(C=C1Br)C#N
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only