missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Bromo-2'-hydroxy-5'-methylacetophenone, 97%, Thermo Scientific Chemicals

$42.55 - $142.62

Chemical Identifiers

CAS 1020253-81-9
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD08273792
InChI Key BPQABGQSVOUODJ-UHFFFAOYSA-N
Synonym 1-4-bromo-2-hydroxy-5-methylphenyl ethanone, 2-acetyl-4-methyl-5-bromophenol, 4'-bromo-2'-hydroxy-5'-methylacetophenone, 1-4-bromo-2-hydroxy-5-methylphenyl ethan-1-one, 1-acetyl-4-bromo-2-hydroxy-5-methylbenzene, 4'-bromo-2'-hydroxy-5'-methyl acetophenone
PubChem CID 42553093
IUPAC Name 1-(4-bromo-2-hydroxy-5-methylphenyl)ethanone
SMILES CC1=C(C=C(C(=C1)C(=O)C)O)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2912503
View Documents
Thermo Scientific Chemicals
H2912503
1 g
Each for $42.55
 
AAH2912506
View Documents
Thermo Scientific Chemicals
H2912506
5 g
Each for $142.62
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1020253-81-9
229.073
BPQABGQSVOUODJ-UHFFFAOYSA-N
42553093
CC1=C(C=C(C(=C1)C(=O)C)O)Br
C9H9BrO2
MFCD08273792
1-4-bromo-2-hydroxy-5-methylphenyl ethanone, 2-acetyl-4-methyl-5-bromophenol, 4'-bromo-2'-hydroxy-5'-methylacetophenone, 1-4-bromo-2-hydroxy-5-methylphenyl ethan-1-one, 1-acetyl-4-bromo-2-hydroxy-5-methylbenzene, 4'-bromo-2'-hydroxy-5'-methyl acetophenone
1-(4-bromo-2-hydroxy-5-methylphenyl)ethanone

Specifications

1020253-81-9
C9H9BrO2
1 g
BPQABGQSVOUODJ-UHFFFAOYSA-N
1-(4-bromo-2-hydroxy-5-methylphenyl)ethanone
42553093
97%
86°C to 89°C
MFCD08273792
1-4-bromo-2-hydroxy-5-methylphenyl ethanone, 2-acetyl-4-methyl-5-bromophenol, 4'-bromo-2'-hydroxy-5'-methylacetophenone, 1-4-bromo-2-hydroxy-5-methylphenyl ethan-1-one, 1-acetyl-4-bromo-2-hydroxy-5-methylbenzene, 4'-bromo-2'-hydroxy-5'-methyl acetophenone
CC1=C(C=C(C(=C1)C(=O)C)O)Br
229.073
229.07
4'-Bromo-2'-hydroxy-5'-methylacetophenone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only