missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-methoxyaniline, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6145406

 View more versions of this product

Catalog No. AAH6145406

Explore available promotions

Description

Description

4-Bromo-2-methoxyaniline is used as a reagent in the synthesis of 3,5-diamino-1,2,4-triazole ureas as potent anaplastic lymphoma kinase (ALK) inhibitors. It is also used as a reagent in the synthesis of chroman-3-amides as potent Rho kinase inhibitors. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-methoxyaniline is used as a reagent in the synthesis of 3,5-diamino-1,2,4-triazole ureas as potent anaplastic lymphoma kinase (ALK) inhibitors. It is also used as a reagent in the synthesis of chroman-3-amides as potent Rho kinase inhibitors. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

4-Bromo-2-methoxyaniline
C7H8BrNO
5 g
4-bromo-2-methoxy-phenylamine, 2-amino-5-bromoanisole, 4-bromo-o-anisidine, 4-bromo-2-methoxyphenylamine, 4-bromo-2-methoxy-aniline, 2-methoxy-4-bromoaniline, benzenamine, 4-bromo-2-methoxy, pubchem2258, acmc-209meb, 4-bromo-2-methoxy aniline
WRFYIYOXJWKONR-UHFFFAOYSA-N
4-bromo-2-methoxyaniline
459257
98%
59557-91-4
MFCD01204266
UN2811
Soluble in methanol.
COC1=C(C=CC(=C1)Br)N
202.051
202.05

Chemical Identifiers

59557-91-4
202.051
WRFYIYOXJWKONR-UHFFFAOYSA-N
459257
COC1=C(C=CC(=C1)Br)N
C7H8BrNO
MFCD01204266
4-bromo-2-methoxy-phenylamine, 2-amino-5-bromoanisole, 4-bromo-o-anisidine, 4-bromo-2-methoxyphenylamine, 4-bromo-2-methoxy-aniline, 2-methoxy-4-bromoaniline, benzenamine, 4-bromo-2-methoxy, pubchem2258, acmc-209meb, 4-bromo-2-methoxy aniline
4-bromo-2-methoxyaniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only