missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-methylanisole, 98+%, Thermo Scientific Chemicals

$79.03 - $275.51

Chemical Identifiers

CAS 14804-31-0
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.063
MDL Number MFCD01321139
InChI Key UDLRGQOHGYWLCS-UHFFFAOYSA-N
Synonym 4-bromo-2-methylanisole, 5-bromo-2-methoxytoluene, benzene, 4-bromo-1-methoxy-2-methyl, 4-bromo-1-methoxy-2-methyl-benzene, 2-methyl-4-bromoanisole, 4-bromo2-methylanisole, acmc-1cfej, 4-bromo2-methyl-anisole, 4-bromo-2-methyl anisole, 4-bromo-2-methyl-anisole
PubChem CID 608315
IUPAC Name 4-bromo-1-methoxy-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Br)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2162109
View Documents
Thermo Scientific Chemicals
B2162109
10 g
Each for $79.03
 
AAB2162118
View Documents
Thermo Scientific Chemicals
B2162118
50 g
Each for $275.51
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14804-31-0
201.063
UDLRGQOHGYWLCS-UHFFFAOYSA-N
608315
CC1=C(C=CC(=C1)Br)OC
C8H9BrO
MFCD01321139
4-bromo-2-methylanisole, 5-bromo-2-methoxytoluene, benzene, 4-bromo-1-methoxy-2-methyl, 4-bromo-1-methoxy-2-methyl-benzene, 2-methyl-4-bromoanisole, 4-bromo2-methylanisole, acmc-1cfej, 4-bromo2-methyl-anisole, 4-bromo-2-methyl anisole, 4-bromo-2-methyl-anisole
4-bromo-1-methoxy-2-methylbenzene

Specifications

14804-31-0
124°C to 125°C (25 mmHg)
MFCD01321139
208224
UDLRGQOHGYWLCS-UHFFFAOYSA-N
4-bromo-1-methoxy-2-methylbenzene
608315
≥98%
66°C to 69°C
C8H9BrO
10 g
4-bromo-2-methylanisole, 5-bromo-2-methoxytoluene, benzene, 4-bromo-1-methoxy-2-methyl, 4-bromo-1-methoxy-2-methyl-benzene, 2-methyl-4-bromoanisole, 4-bromo2-methylanisole, acmc-1cfej, 4-bromo2-methyl-anisole, 4-bromo-2-methyl anisole, 4-bromo-2-methyl-anisole
CC1=C(C=CC(=C1)Br)OC
201.063
201.06
4-Bromo-2-methylanisole
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only