missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-methylphenol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3230714

 View more versions of this product

Catalog No. AAH3230714


Explore available promotions

Description

Description

4-Bromo-2-methylphenol, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-methylphenol, is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

4-Bromo-2-methylphenol
62°C to 66°C
C13H9F3O
25 g
Slightly soluble in water.
C1=CC=C(C=C1)OC2=CC=C(C=C2)C(F)(F)F
238.21
187.04
2362-12-1
Phenol-like
MFCD00055435
4-trifluoromethyl diphenyl ether, 1-phenoxy-4-trifluoromethyl benzene, 4-phenoxy-1-trifluoromethyl benzene, 4-phenoxybenzotrifluoride, trifluoromethyldiphenyl ether, 4-trifluoromethyl diphenylether, 4-phenoxy-alpha,alpha,alpha-trifluorotoluene
UZJUDUZMPNCXPF-UHFFFAOYSA-N
1-phenoxy-4-(trifluoromethyl)benzene
2777517
97%

Chemical Identifiers

2362-12-1
MFCD00055435
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H319-H335

RTECSNumber : GO6868000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only