missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-(trifluoromethoxy)aniline 98.0+%, TCI America™
$41.02 - $108.11
Chemical Identifiers
| CAS | 175278-09-8 |
|---|---|
| Molecular Formula | C7H5BrF3NO |
| Molecular Weight (g/mol) | 256.02 |
| MDL Number | MFCD00179338 |
| InChI Key | QVILSWLYJYMGRN-UHFFFAOYSA-N |
| Synonym | 4-bromo-2-trifluoromethoxy aniline, 4-bromo-2-trifluoromethoxy-phenylamine, 2-amino-5-bromo trifluoromethoxy benzene, benzenamine, 4-bromo-2-trifluoromethoxy, 1-amino-4-bromo-2-trifluoromethoxy benzene, buttpark 50\01-74, 4-bromo-2-trifluoromethoxy phenylamine, pubchem2930, acmc-1c9j8, ksc184e5f |
| PubChem CID | 737382 |
| IUPAC Name | 4-bromo-2-(trifluoromethoxy)aniline |
| SMILES | NC1=CC=C(Br)C=C1OC(F)(F)F |
Chemical Identifiers
| 175278-09-8 | |
| 256.02 | |
| QVILSWLYJYMGRN-UHFFFAOYSA-N | |
| 737382 | |
| NC1=CC=C(Br)C=C1OC(F)(F)F |
| C7H5BrF3NO | |
| MFCD00179338 | |
| 4-bromo-2-trifluoromethoxy aniline, 4-bromo-2-trifluoromethoxy-phenylamine, 2-amino-5-bromo trifluoromethoxy benzene, benzenamine, 4-bromo-2-trifluoromethoxy, 1-amino-4-bromo-2-trifluoromethoxy benzene, buttpark 50\01-74, 4-bromo-2-trifluoromethoxy phenylamine, pubchem2930, acmc-1c9j8, ksc184e5f | |
| 4-bromo-2-(trifluoromethoxy)aniline |
Specifications
| 175278-09-8 | |
| 115°C | |
| MFCD00179338 | |
| 4-bromo-2-trifluoromethoxy aniline, 4-bromo-2-trifluoromethoxy-phenylamine, 2-amino-5-bromo trifluoromethoxy benzene, benzenamine, 4-bromo-2-trifluoromethoxy, 1-amino-4-bromo-2-trifluoromethoxy benzene, buttpark 50\01-74, 4-bromo-2-trifluoromethoxy phenylamine, pubchem2930, acmc-1c9j8, ksc184e5f | |
| NC1=CC=C(Br)C=C1OC(F)(F)F | |
| 256.02 | |
| 256.02 | |
| Liquid |
| Yellow | |
| C7H5BrF3NO | |
| 5 g | |
| QVILSWLYJYMGRN-UHFFFAOYSA-N | |
| 4-bromo-2-(trifluoromethoxy)aniline | |
| 737382 | |
| ≥98.0% (GC) | |
| 4-Bromo-2-(trifluoromethoxy)aniline |
Safety and Handling
TSCA : No