missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-3,5-dimethylphenol, 99%, Thermo Scientific Chemicals

$100.44 - $323.23

Chemical Identifiers

CAS 7463-51-6
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.063
MDL Number MFCD00002315
InChI Key WMUWDPLTTLJNPE-UHFFFAOYSA-N
Synonym 4-bromo-3,5-xylenol, 4-bromo-3,5-dimethyl phenol, phenol, 4-bromo-3,5-dimethyl, 3,5-dimethyl-4-bromophenol, 4-bromo-3,5-dimethyl-phenol, zlchem 241, pubchem3767, acmc-209ovl, 3,5-xylenol, 4-bromo, akos bbb/606
PubChem CID 81970
IUPAC Name 4-bromo-3,5-dimethylphenol
SMILES CC1=CC(=CC(=C1Br)C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2168406
View Documents
Thermo Scientific Chemicals
B2168406
5 g
Each for $100.44
 
AAB2168414
View Documents
Thermo Scientific Chemicals
B2168414
25 g
Each for $323.23
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7463-51-6
201.063
WMUWDPLTTLJNPE-UHFFFAOYSA-N
81970
CC1=CC(=CC(=C1Br)C)O
C8H9BrO
MFCD00002315
4-bromo-3,5-xylenol, 4-bromo-3,5-dimethyl phenol, phenol, 4-bromo-3,5-dimethyl, 3,5-dimethyl-4-bromophenol, 4-bromo-3,5-dimethyl-phenol, zlchem 241, pubchem3767, acmc-209ovl, 3,5-xylenol, 4-bromo, akos bbb/606
4-bromo-3,5-dimethylphenol

Specifications

7463-51-6
Phenol-like
MFCD00002315
2207127
WMUWDPLTTLJNPE-UHFFFAOYSA-N
4-bromo-3,5-dimethylphenol
81970
99%
113°C to 117°C
C8H9BrO
5 g
4-bromo-3,5-xylenol, 4-bromo-3,5-dimethyl phenol, phenol, 4-bromo-3,5-dimethyl, 3,5-dimethyl-4-bromophenol, 4-bromo-3,5-dimethyl-phenol, zlchem 241, pubchem3767, acmc-209ovl, 3,5-xylenol, 4-bromo, akos bbb/606
CC1=CC(=CC(=C1Br)C)O
201.063
201.07
4-Bromo-3,5-dimethylphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335-H500

EINECSNumber : 231-255-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only