Learn More
4-Bromo-3-chlorophenol, 98%, Thermo Scientific Chemicals
$186.27 - $186.27
Chemical Identifiers
CAS | 13631-21-5 |
---|---|
Molecular Formula | C6H4BrClO |
Molecular Weight (g/mol) | 207.451 |
MDL Number | MFCD00070744 |
InChI Key | FQEYHIPPYOSPLF-UHFFFAOYSA-N |
Synonym | phenol, 4-bromo-3-chloro, 3-chloro-4-bromophenol, 2-chloro-4-hydroxybromobenzene, pubchem3597, acmc-1bvpq, 4-bromo-3-chloro-phenol, intermediates-zcf02051, ksc493i5n, 4-bromanyl-3-chloranyl-phenol |
PubChem CID | 7018182 |
IUPAC Name | 4-bromo-3-chlorophenol |
SMILES | C1=CC(=C(C=C1O)Cl)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL1380106
|
Thermo Scientific Chemicals
L1380106 |
5 g |
Each for $186.27
|
|
||||||
Description
4-Bromo-3-chlorophenol is used as a precursor in organic synthesis to prepare 4-(benzyloxy)-l-bromo-2-chlorobenzene by reaction with benzyl bromide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromo-3-chlorophenol is used as a precursor in organic synthesis to prepare 4-(benzyloxy)-l-bromo-2-chlorobenzene by reaction with benzyl bromide.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
13631-21-5 | |
207.451 | |
FQEYHIPPYOSPLF-UHFFFAOYSA-N | |
7018182 | |
C1=CC(=C(C=C1O)Cl)Br |
C6H4BrClO | |
MFCD00070744 | |
phenol, 4-bromo-3-chloro, 3-chloro-4-bromophenol, 2-chloro-4-hydroxybromobenzene, pubchem3597, acmc-1bvpq, 4-bromo-3-chloro-phenol, intermediates-zcf02051, ksc493i5n, 4-bromanyl-3-chloranyl-phenol | |
4-bromo-3-chlorophenol |
Specifications
13631-21-5 | |
Phenol-like | |
MFCD00070744 | |
3236445 | |
Slightly soluble in water. | |
C1=CC(=C(C=C1O)Cl)Br | |
207.451 | |
207.46 | |
4-Bromo-3-chlorophenol |
64°C to 68°C | |
C6H4BrClO | |
5 g | |
phenol, 4-bromo-3-chloro, 3-chloro-4-bromophenol, 2-chloro-4-hydroxybromobenzene, pubchem3597, acmc-1bvpq, 4-bromo-3-chloro-phenol, intermediates-zcf02051, ksc493i5n, 4-bromanyl-3-chloranyl-phenol | |
FQEYHIPPYOSPLF-UHFFFAOYSA-N | |
4-bromo-3-chlorophenol | |
7018182 | |
98% |
Safety and Handling
GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only