missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-3-methylaniline, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 167502500

 View more versions of this product

Catalog No. AC167502500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

4-Bromo-3-methylaniline
6933-10-4
100.0
96% min. (GC)
BrC6H3(CH3)NH2
250g
methyl 2-aminoisonicotinate, 2-aminopyridine-4-carboxylic acid methyl ester, 2-aminoisonicotinic acid methyl ester, 4-methoxycarbonyl pyridin-2-amine, 4-pyridinecarboxylic acid, 2-amino-, methyl ester, methyl 2-amino-4-pyridinecarboxylate, methyl-2-aminopyridine-4-carboxylate, 2-amino-4-pyridinecarboxylic acid methylester, 2-amino-4-pyridinecarboxylic acid methyl ester, methyl-2-aminoisonicotinat
SVWWNEYBEFASMP-UHFFFAOYSA-N
methyl 2-aminopyridine-4-carboxylate
236984
97%
99%
98.5
240.0°C
C7H8BrN
MFCD00007828
12, 873
Solubility in water: insoluble
COC(=O)C1=CC(=NC=C1)N
186.05
186.05

Chemical Identifiers

6933-10-4
186.05
SVWWNEYBEFASMP-UHFFFAOYSA-N
236984
COC(=O)C1=CC(=NC=C1)N
C7H8BrN
MFCD00007828
methyl 2-aminoisonicotinate, 2-aminopyridine-4-carboxylic acid methyl ester, 2-aminoisonicotinic acid methyl ester, 4-methoxycarbonyl pyridin-2-amine, 4-pyridinecarboxylic acid, 2-amino-, methyl ester, methyl 2-amino-4-pyridinecarboxylate, methyl-2-aminopyridine-4-carboxylate, 2-amino-4-pyridinecarboxylic acid methylester, 2-amino-4-pyridinecarboxylic acid methyl ester, methyl-2-aminoisonicotinat
methyl 2-aminopyridine-4-carboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 230-056-3

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only