missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Bromo-N-(4-fluorophenyl)benzamide, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H5740903
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Bromo-N-(4-fluorophenyl)benzamide | |
C13H9BrFNO | |
1 g | |
DBCKBDOQSKIIOA-UHFFFAOYSA-N | |
4-bromo-N-(4-fluorophenyl)benzamide | |
532150 | |
97% |
346723-41-9 | |
MFCD00442009 | |
4-bromo-n-4-fluorophenyl benzamide, benzamide, n-4-fluorophenyl-4-bromo, cambridge id 5270654, n-4-fluorophenyl-4-bromobenzamide, 4-bromo-n-4-fluoro-phenyl-benzamide, 4-bromophenyl-n-4-fluorophenyl carboxamide | |
C1=CC(=CC=C1C(=O)NC2=CC=C(C=C2)F)Br | |
294.123 | |
294.12 |
Chemical Identifiers
346723-41-9 | |
294.123 | |
DBCKBDOQSKIIOA-UHFFFAOYSA-N | |
532150 | |
C1=CC(=CC=C1C(=O)NC2=CC=C(C=C2)F)Br |
C13H9BrFNO | |
MFCD00442009 | |
4-bromo-n-4-fluorophenyl benzamide, benzamide, n-4-fluorophenyl-4-bromo, cambridge id 5270654, n-4-fluorophenyl-4-bromobenzamide, 4-bromo-n-4-fluoro-phenyl-benzamide, 4-bromophenyl-n-4-fluorophenyl carboxamide | |
4-bromo-N-(4-fluorophenyl)benzamide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures