missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-N-(4-fluorophenyl)benzamide, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H57409MD

 View more versions of this product

Catalog No. AAH57409MD


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-Bromo-N-(4-fluorophenyl)benzamide
C13H9BrFNO
250 mg
DBCKBDOQSKIIOA-UHFFFAOYSA-N
4-bromo-N-(4-fluorophenyl)benzamide
532150
97%
346723-41-9
MFCD00442009
4-bromo-n-4-fluorophenyl benzamide, benzamide, n-4-fluorophenyl-4-bromo, cambridge id 5270654, n-4-fluorophenyl-4-bromobenzamide, 4-bromo-n-4-fluoro-phenyl-benzamide, 4-bromophenyl-n-4-fluorophenyl carboxamide
C1=CC(=CC=C1C(=O)NC2=CC=C(C=C2)F)Br
294.123
294.12

Chemical Identifiers

346723-41-9
294.123
DBCKBDOQSKIIOA-UHFFFAOYSA-N
532150
C1=CC(=CC=C1C(=O)NC2=CC=C(C=C2)F)Br
C13H9BrFNO
MFCD00442009
4-bromo-n-4-fluorophenyl benzamide, benzamide, n-4-fluorophenyl-4-bromo, cambridge id 5270654, n-4-fluorophenyl-4-bromobenzamide, 4-bromo-n-4-fluoro-phenyl-benzamide, 4-bromophenyl-n-4-fluorophenyl carboxamide
4-bromo-N-(4-fluorophenyl)benzamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions