missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-N,N,2,6-tetramethylaniline, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5058406

 View more versions of this product

Catalog No. AAH5058406

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromo-N,N,2,6-tetramethylaniline
5 g
MFCD00100113
NDDKEDQKCHIHMI-UHFFFAOYSA-N
4-bromo-N,N,2,6-tetramethylaniline
142731
98%
50638-54-5
C10H14BrN
n1,n1,2,6-tetramethyl-4-bromoaniline, n-4-bromo-2,6-dimethylphenyl-n,n-dimethylamine, n,n,2,6-tetramethylaniline, 4-bromo, 4-bromo-2,6-dimethylphenyl dimethylamine, pubchem5369, acmc-20ak7q, n,n,2,6-tetramethylaniline,4-bromo, 4-bromanyl-n,n,2,6-tetramethyl-aniline, 4-bromo-n,n-dimethyl-2,6-xylidine, benzenamine,4-bromo-n,n,2,6-tetramethyl
CC1=CC(=CC(=C1N(C)C)C)Br
228.133
228.13

Chemical Identifiers

50638-54-5
228.133
NDDKEDQKCHIHMI-UHFFFAOYSA-N
4-bromo-N,N,2,6-tetramethylaniline
C10H14BrN
MFCD00100113
142731
CC1=CC(=CC(=C1N(C)C)C)Br
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only