missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Bromoacetophenone, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 106591000

 View more versions of this product

Catalog No. AC106591000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4'-Bromoacetophenone
97.5
48°C to 52°C
255°C
Authentic
Glass bottle
BrC6H4COCH3
100 g
15,141
Solubility in water: insoluble
CC(=O)C1=CC=C(Br)C=C1
199.05
199.05
Crystalline Flakes or Crystals
99-90-1
100.0
White to Yellow
>100°C
97.5% min. (GC)
C8H7BrO
MFCD00000105
07,283
4'-bromoacetophenone, 1-4-bromophenyl ethanone, p-bromoacetophenone, 4-bromoacetophenone, ethanone, 1-4-bromophenyl, 1-4-bromophenyl ethan-1-one, 1-acetyl-4-bromobenzene, methyl p-bromophenyl ketone, p-bromophenyl methyl ketone, acetophenone, 4'-bromo
WYECURVXVYPVAT-UHFFFAOYSA-N
1-(4-bromophenyl)ethanone
7466
98%

Chemical Identifiers

99-90-1
199.05
WYECURVXVYPVAT-UHFFFAOYSA-N
7466
CC(=O)C1=CC=C(Br)C=C1
C8H7BrO
MFCD00000105
4'-bromoacetophenone, 1-4-bromophenyl ethanone, p-bromoacetophenone, 4-bromoacetophenone, ethanone, 1-4-bromophenyl, 1-4-bromophenyl ethan-1-one, 1-acetyl-4-bromobenzene, methyl p-bromophenyl ketone, p-bromophenyl methyl ketone, acetophenone, 4'-bromo
1-(4-bromophenyl)ethanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 202-799-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only