Learn More
4-(Bromoacetyl)pyridine hydrobromide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3228503
Description
Design and synthesis of potent, selective, and orally bioavailable tetrasubstituted imidazole inhibitors of p38 mitogen-activated protein kinase 4-(Bromoacetyl)pyridine hydrobromide was used to prepare 2-Phenyl-4-(4-pyridyl)imidazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDesign and synthesis of potent, selective, and orally bioavailable tetrasubstituted imidazole inhibitors of p38 mitogen-activated protein kinase 4-(Bromoacetyl)pyridine hydrobromide was used to prepare 2-Phenyl-4-(4-pyridyl)imidazole.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
Specifications
4-(Bromoacetyl)pyridine hydrobromide | |
∽200°C (decomposition) | |
C7H7Br2NO | |
1 g | |
4-bromoacetyl pyridine hydrobromide, 2-bromo-1-pyridin-4-yl ethanone hydrobromide, 2-bromo-1-4-pyridinyl-1-ethanone hydrobromide, 2-bromo-1-pyridin-4-yl ethan-1-one hydrobromide, ethanone, 2-bromo-1-4-pyridinyl-, hydrobromide, 4-2-bromoacetyl pyridine hydrobromide, 4-bromoacetylpyridine hydrobromide, 2-bromo-1-pyrdin-4-ylethan-1-one hydrobromide, 2-bromo-1-4-pyridinyl-1-ethanone hbr, 4-bromoacetyl pyridine hbr | |
RGALBQILADNMKA-UHFFFAOYSA-N | |
2-bromo-1-pyridin-4-ylethanone;hydrobromide | |
2776239 | |
98% |
5349-17-7 | |
Weak | |
MFCD02681893 | |
UN3261 | |
Soluble in water. | |
C1=CN=CC=C1C(=O)CBr.Br | |
280.947 | |
280.95 |
Chemical Identifiers
5349-17-7 | |
280.947 | |
RGALBQILADNMKA-UHFFFAOYSA-N | |
2776239 | |
C1=CN=CC=C1C(=O)CBr.Br |
C7H7Br2NO | |
MFCD02681893 | |
4-bromoacetyl pyridine hydrobromide, 2-bromo-1-pyridin-4-yl ethanone hydrobromide, 2-bromo-1-4-pyridinyl-1-ethanone hydrobromide, 2-bromo-1-pyridin-4-yl ethan-1-one hydrobromide, ethanone, 2-bromo-1-4-pyridinyl-, hydrobromide, 4-2-bromoacetyl pyridine hydrobromide, 4-bromoacetylpyridine hydrobromide, 2-bromo-1-pyrdin-4-ylethan-1-one hydrobromide, 2-bromo-1-4-pyridinyl-1-ethanone hbr, 4-bromoacetyl pyridine hbr | |
2-bromo-1-pyridin-4-ylethanone;hydrobromide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only