Learn More
4-Bromoaniline, 99+%, Thermo Scientific Chemicals
$190.49 - $597.85
Chemical Identifiers
CAS | 106-40-1 |
---|---|
Molecular Formula | C6H6BrN |
Molecular Weight (g/mol) | 172.02 |
InChI Key | WDFQBORIUYODSI-UHFFFAOYSA-N |
Synonym | p-bromoaniline, 4-bromobenzenamine, benzenamine, 4-bromo, p-bromophenylamine, aniline, p-bromo, 4-bromo-aniline, 4-bromobenzeneamine, p-bromo aniline, 4-bromanilinu, 4-bromo-phenylamine |
PubChem CID | 7807 |
IUPAC Name | 4-bromoaniline |
SMILES | C1=CC(=CC=C1N)Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC146280250
|
Thermo Scientific Chemicals
146280250 |
25 g | Glass bottle |
Each for $190.49
|
|
|||||
AC146281000
|
Thermo Scientific Chemicals
146281000 |
100 g | Glass bottle |
Each for $597.85
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
106-40-1 | |
172.02 | |
p-bromoaniline, 4-bromobenzenamine, benzenamine, 4-bromo, p-bromophenylamine, aniline, p-bromo, 4-bromo-aniline, 4-bromobenzeneamine, p-bromo aniline, 4-bromanilinu, 4-bromo-phenylamine | |
4-bromoaniline |
C6H6BrN | |
WDFQBORIUYODSI-UHFFFAOYSA-N | |
7807 | |
C1=CC(=CC=C1N)Br |
Specifications
106-40-1 | |
Brown-Gray to White | |
230.0°C to 250.0°C | |
Authentic | |
Glass bottle | |
BrC6H4NH2 | |
12,636 | |
15,1411 | |
Solubility in water: < 1 g / l. Other solubilities: soluble in most organic solvents except cold,petroleum,very soluble in alcohol and ether | |
C1=CC(=CC=C1N)Br | |
172.02 | |
172.02 | |
Crystalline Powder |
60.0°C to 64.0°C | |
1.4970g/mL | |
>110°C | |
99% min. (GC) | |
C6H6BrN | |
25 g | |
1.497 | |
p-bromoaniline, 4-bromobenzenamine, benzenamine, 4-bromo, p-bromophenylamine, aniline, p-bromo, 4-bromo-aniline, 4-bromobenzeneamine, p-bromo aniline, 4-bromanilinu, 4-bromo-phenylamine | |
WDFQBORIUYODSI-UHFFFAOYSA-N | |
4-bromoaniline | |
7807 | |
99+% | |
4-Bromoaniline |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
GHS Signal Word: Danger
EINECSNumber : 203-393-9
RUO – Research Use Only