missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromoanisole, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1182436

 View more versions of this product

Catalog No. AAA1182436


Explore available promotions

Description

Description

4-Bromoanisole finds application as an intermediate in synthetic chemistry. It is used in the preparation of aryl 1,3-diketones and ethyl 4-methoxycinnamate. It is a used as a brominating reagent. Further, it is used in Suzuki coupling reaction with phenylboronic acid as well as in Heck reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromoanisole finds application as an intermediate in synthetic chemistry. It is used in the preparation of aryl 1,3-diketones and ethyl 4-methoxycinnamate. It is a used as a brominating reagent. Further, it is used in Suzuki coupling reaction with phenylboronic acid as well as in Heck reaction.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications

Specifications

4-Bromoanisole
10°C to 13°C
98°C to 99°C (10 mmHg)
99%
1.564
500 g
14,1427
Immiscible with water.
COC1=CC=C(C=C1)Br
187.036
CHEBI:47257
99%
104-92-7
1.494
94°C (201°F)
C7H7BrO
MFCD00000097
1237590
4-bromoanisole, p-bromoanisole, benzene, 1-bromo-4-methoxy, 4-methoxybromobenzene, anisyl bromide, p-anisyl bromide, p-bromanisole, p-methoxybromobenzene, p-methoxyphenyl bromide, 4-methoxyphenyl bromide
QJPJQTDYNZXKQF-UHFFFAOYSA-N
1-bromo-4-methoxybenzene
7730
187.04
Liquid

Chemical Identifiers

104-92-7
187.036
QJPJQTDYNZXKQF-UHFFFAOYSA-N
7730
1-bromo-4-methoxybenzene
C7H7BrO
MFCD00000097
4-bromoanisole, p-bromoanisole, benzene, 1-bromo-4-methoxy, 4-methoxybromobenzene, anisyl bromide, p-anisyl bromide, p-bromanisole, p-methoxybromobenzene, p-methoxyphenyl bromide, 4-methoxyphenyl bromide
CHEBI:47257
COC1=CC=C(C=C1)Br
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-252-1

RTECSNumber : BZ8501000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only