missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromobenz[a]anthracene 97.0+%, TCI America™
$48.46 - $137.94
Chemical Identifiers
| CAS | 61921-39-9 |
|---|---|
| Molecular Formula | C18H11Br |
| Molecular Weight (g/mol) | 307.19 |
| MDL Number | MFCD25371993 |
| InChI Key | ILVKEMDEUCXCAB-UHFFFAOYSA-N |
| Synonym | 4-Bromotetraphene |
| PubChem CID | 182324 |
| IUPAC Name | 4-bromobenzo[a]anthracene |
| SMILES | C1=CC=C2C=C3C(=CC2=C1)C=CC4=C3C=CC=C4Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B5450200MG
|
TCI America
B5450200MG |
200 mg |
Each for $48.46
|
|
|||||
|
B54501G
|
TCI America
B54501G |
1 g |
Each for $137.94
|
|
|||||
Chemical Identifiers
| 61921-39-9 | |
| 307.19 | |
| ILVKEMDEUCXCAB-UHFFFAOYSA-N | |
| 182324 | |
| C1=CC=C2C=C3C(=CC2=C1)C=CC4=C3C=CC=C4Br |
| C18H11Br | |
| MFCD25371993 | |
| 4-Bromotetraphene | |
| 4-bromobenzo[a]anthracene |
Specifications
| 61921-39-9 | |
| C18H11Br | |
| 200 mg | |
| ILVKEMDEUCXCAB-UHFFFAOYSA-N | |
| 4-bromobenzo[a]anthracene | |
| 182324 | |
| ≥97.0% (GC) | |
| 4-Bromobenz[a]anthracene |
| 220°C | |
| MFCD25371993 | |
| 4-Bromotetraphene | |
| C1=CC=C2C=C3C(=CC2=C1)C=CC4=C3C=CC=C4Br | |
| 307.19 | |
| 307.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No