missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzoyl chloride, 98+%

CAS: 586-75-4 | C7H4BrClO | 219.46 g/mol

$76.07 - $307.16

Chemical Identifiers

CAS 586-75-4
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.46
MDL Number MFCD00000683
InChI Key DENKGPBHLYFNGK-UHFFFAOYSA-N
Synonym p-bromobenzoyl chloride, benzoyl chloride, 4-bromo, benzoyl chloride, p-bromo, 4-bromobenzoylchloride, p-bromobenzoyl chloride, p-bromo, 4-bromobenzoic acid chloride, p-brombenzoylchlorid, 4bromobenzoyl chloride, p-brc6h4cocl, 4-bromo-benzoylchloride
PubChem CID 68515
IUPAC Name 4-bromobenzoyl chloride
SMILES ClC(=O)C1=CC=C(Br)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0379409
View Documents
Thermo Scientific Chemicals
L0379409
10 g
Each for $76.07
Only null left
 
AAL0379418
View Documents
Thermo Scientific Chemicals
L0379418
50 g
Each for $307.16
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Bromobenzoyl chloride is used in the synthesis of HIV-1 protease inhibitors. It is also used in the synthesis of oxazoles, cyclohexanone derivatives as analgesic and antimicrobial agents.

Solubility
Soluble in methanol. Reacts with water forming Hcl.

Notes
Moisture Sensitive. Store away from strong bases. Incompatible with moisture and bases. Store at 2 - 8°C.
Specifications

Chemical Identifiers

586-75-4
219.46
DENKGPBHLYFNGK-UHFFFAOYSA-N
68515
ClC(=O)C1=CC=C(Br)C=C1
C7H4BrClO
MFCD00000683
p-bromobenzoyl chloride, benzoyl chloride, 4-bromo, benzoyl chloride, p-bromo, 4-bromobenzoylchloride, p-bromobenzoyl chloride, p-bromo, 4-bromobenzoic acid chloride, p-brombenzoylchlorid, 4bromobenzoyl chloride, p-brc6h4cocl, 4-bromo-benzoylchloride
4-bromobenzoyl chloride

Specifications

586-75-4
246°C
C7H4BrClO
10 g
636641
p-bromobenzoyl chloride, benzoyl chloride, 4-bromo, benzoyl chloride, p-bromo, 4-bromobenzoylchloride, p-bromobenzoyl chloride, p-bromo, 4-bromobenzoic acid chloride, p-brombenzoylchlorid, 4bromobenzoyl chloride, p-brc6h4cocl, 4-bromo-benzoylchloride
DENKGPBHLYFNGK-UHFFFAOYSA-N
4-bromobenzoyl chloride
68515
≥98%
38°C to 42°C
>110°C (230°F)
MFCD00000683
UN3261
Moisture sensitive
Soluble in methanol. Reacts with water forming Hcl.
ClC(=O)C1=CC=C(Br)C=C1
219.46
219.47
4-Bromobenzoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 209-580-1

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only