missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromobiphenyl, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 197981000

 View more versions of this product

Catalog No. AC197981000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromobiphenyl
92-66-0
100.0
White to Yellow
143°C
99%
C12H9Br
MFCD00000100
05, 580
Solubility in water: insoluble. Other solubilities: soluble in ethanol, ether and benzene
C1=CC=C(C=C1)C2=CC=C(C=C2)Br
233.11
233.11
Crystalline Flakes
99%
98.5
89.0°C to 92.0°C
310.0°C
Authentic
Glass bottle
C6H5C6H4Br
100 g
4-bromobiphenyl, p-bromobiphenyl, 4-bromo-1,1'-biphenyl, 4-bromodiphenyl, 1,1'-biphenyl, 4-bromo, 4-biphenyl bromide, p-bromodiphenyl, biphenyl, 4-bromo, 4-bromo-biphenyl, p-phenylbromobenzene
PKJBWOWQJHHAHG-UHFFFAOYSA-N
1-bromo-4-phenylbenzene
7101
99%

Chemical Identifiers

92-66-0
233.11
PKJBWOWQJHHAHG-UHFFFAOYSA-N
7101
C1=CC=C(C=C1)C2=CC=C(C=C2)Br
C12H9Br
MFCD00000100
4-bromobiphenyl, p-bromobiphenyl, 4-bromo-1,1'-biphenyl, 4-bromodiphenyl, 1,1'-biphenyl, 4-bromo, 4-biphenyl bromide, p-bromodiphenyl, biphenyl, 4-bromo, 4-bromo-biphenyl, p-phenylbromobenzene
1-bromo-4-phenylbenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Harmful if swallowed.
Very toxic to aquatic life.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

EINECSNumber : 202-176-6

RTECSNumber : DV1750100

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only