Learn More
4-Bromobutyric acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1287009
Description
4-Bromobutyric acid is used as a bifunctional cross-linker and as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromobutyric acid is used as a bifunctional cross-linker and as a pharmaceutical intermediate.
Solubility
Soluble in water and dilute alkali.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at room temperature. It is sensitive to light. Incompatible with strong oxidizing agents
Specifications
4-Bromobutyric acid | |
31°C to 33°C | |
115°C (239°F) | |
C4H7BrO2 | |
10 g | |
1743056 | |
4-bromobutyric acid, butanoic acid, 4-bromo, 4-bromo-n-butyric acid, butyric acid, 4-bromo, gamma-bromobutyric acid, rarechem al bo 0176, 4-bromobutyricacid, carboxypropyl bromide, 4-bromo-butyric acid, pubchem3754 | |
GRHQDJDRGZFIPO-UHFFFAOYSA-N | |
4-bromobutanoic acid | |
75809 | |
97% |
2623-87-2 | |
129°C to 131°C (11 mmHg) | |
Stench | |
MFCD00002817 | |
UN3261 | |
Light sensitive | |
Soluble in water and dilute alkali. | |
OC(=O)CCCBr | |
167.00 | |
167.01 |
Chemical Identifiers
2623-87-2 | |
167.00 | |
GRHQDJDRGZFIPO-UHFFFAOYSA-N | |
75809 | |
OC(=O)CCCBr |
C4H7BrO2 | |
MFCD00002817 | |
4-bromobutyric acid, butanoic acid, 4-bromo, 4-bromo-n-butyric acid, butyric acid, 4-bromo, gamma-bromobutyric acid, rarechem al bo 0176, 4-bromobutyricacid, carboxypropyl bromide, 4-bromo-butyric acid, pubchem3754 | |
4-bromobutanoic acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 220-083-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only