missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromobutyric acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 133470050

 View more versions of this product

Catalog No. AC133470050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromobutyric acid
2623-87-2
100.0
Brown
124°C
98%
C4H7BrO2
MFCD00002817
02, 283
Solubility in water: .. Other solubilities: soluble in dilute alkali,soluble in most common organic solvents
OC(=O)CCCBr
167.00
167
Low Melting Crystals
98%
97.5
28°C to 32°C
301°C
Authentic
Glass Bottle
Br(CH2)3COOH
5 g
4-bromobutyric acid, butanoic acid, 4-bromo, 4-bromo-n-butyric acid, butyric acid, 4-bromo, gamma-bromobutyric acid, rarechem al bo 0176, 4-bromobutyricacid, carboxypropyl bromide, 4-bromo-butyric acid, pubchem3754
GRHQDJDRGZFIPO-UHFFFAOYSA-N
4-bromobutanoic acid
75809
98%

Chemical Identifiers

2623-87-2
167.00
GRHQDJDRGZFIPO-UHFFFAOYSA-N
75809
OC(=O)CCCBr
C4H7BrO2
MFCD00002817
4-bromobutyric acid, butanoic acid, 4-bromo, 4-bromo-n-butyric acid, butyric acid, 4-bromo, gamma-bromobutyric acid, rarechem al bo 0176, 4-bromobutyricacid, carboxypropyl bromide, 4-bromo-butyric acid, pubchem3754
4-bromobutanoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 220-083-9

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only