missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromochlorobenzene, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 106835000

 View more versions of this product

Catalog No. AC106835000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromochlorobenzene
99%
White
100.0
196.0°C
Authentic
Crystalline Powder
1.651
BrC6H4Cl
05, 209
Solubility in water: 0.045g/L (25°C). Other solubilities: soluble in most organic solvents
ClC1=CC=C(Br)C=C1
191.45
191.45
64.0°C to 67.0°C
106-39-8
98.5
1.6510g/mL
500 g
Plastic bottle
98.5% min. (GC)
C6H4BrCl
MFCD00000600
4-bromochlorobenzene, p-bromochlorobenzene, 4-chlorobromobenzene, p-chlorobromobenzene, p-chlorophenyl bromide, benzene, 1-bromo-4-chloro, 4-chlorophenyl bromide, 1-chloro-4-bromobenzene, 4-chloro-1-bromobenzene, p-bromoclorobenzene
NHDODQWIKUYWMW-UHFFFAOYSA-N
1-bromo-4-chlorobenzene
7806
99%

Chemical Identifiers

106-39-8
191.45
NHDODQWIKUYWMW-UHFFFAOYSA-N
7806
ClC1=CC=C(Br)C=C1
C6H4BrCl
MFCD00000600
4-bromochlorobenzene, p-bromochlorobenzene, 4-chlorobromobenzene, p-chlorobromobenzene, p-chlorophenyl bromide, benzene, 1-bromo-4-chloro, 4-chlorophenyl bromide, 1-chloro-4-bromobenzene, 4-chloro-1-bromobenzene, p-bromoclorobenzene
1-bromo-4-chlorobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-392-3

RTECSNumber : CY9006000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only