missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromofluorobenzene, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 107015000

 View more versions of this product

Catalog No. AC107015000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromofluorobenzene
460-00-4
100.0
Colorless
150.0°C to 153.0°C
Authentic
Glass bottle
1.5260 to 1.5280
MFCD00000342
05, 209
4-bromofluorobenzene, p-bromofluorobenzene, p-fluorobromobenzene, 4-fluorobromobenzene, benzene, 1-bromo-4-fluoro, 4-fluorophenyl bromide, 1-fluoro-4-bromobenzene, 4-fluoro-1-bromobenzene, p-fluorophenyl bromide, bromo fluorobenzene
AITNMTXHTIIIBB-UHFFFAOYSA-N
1-bromo-4-fluorobenzene
9993
99%
99%
98.5
-8.0°C
1.5930g/mL
53°C
99%
C6H4BrF
BrC6H4F
500 mL
1.593
Solubility in water: 0.14g/L (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,acetone,soluble in ether
FC1=CC=C(Br)C=C1
175.00
175
Liquid

Chemical Identifiers

460-00-4
175.00
AITNMTXHTIIIBB-UHFFFAOYSA-N
9993
FC1=CC=C(Br)C=C1
C6H4BrF
MFCD00000342
4-bromofluorobenzene, p-bromofluorobenzene, p-fluorobromobenzene, 4-fluorobromobenzene, benzene, 1-bromo-4-fluoro, 4-fluorophenyl bromide, 1-fluoro-4-bromobenzene, 4-fluoro-1-bromobenzene, p-fluorophenyl bromide, bromo fluorobenzene
1-bromo-4-fluorobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.

GHS Signal Word: Warning

EINECSNumber : 207-300-2

RTECSNumber : CY9033750

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only