Learn More
4-Bromoheptane, 97%, Thermo Scientific Chemicals
$108.66 - $321.41
Chemical Identifiers
CAS | 998-93-6 |
---|---|
Molecular Formula | C7H15Br |
Molecular Weight (g/mol) | 179.10 |
MDL Number | MFCD00039182 |
InChI Key | BNUTXEKPXPZAIT-UHFFFAOYSA-N |
Synonym | heptane, 4-bromo, 4-bromo-heptane, acmc-20ammr, 4-bromoheptane 5g |
PubChem CID | 70460 |
IUPAC Name | 4-bromoheptane |
SMILES | CCCC(Br)CCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0480706
|
Thermo Scientific Chemicals
L0480706 |
5 g |
Each for $108.66
|
|
|||||
AAL0480714
|
Thermo Scientific Chemicals
L0480714 |
25 g |
Each for $321.41
|
|
|||||
Description
4-Bromoheptane is used in the preparation of 4-nitro-heptane by reacting with sodium nitrite as a reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromoheptane is used in the preparation of 4-nitro-heptane by reacting with sodium nitrite as a reagent.
Solubility
Miscible with carbon tetrachloride, chloroform and benzene. Immiscible with water.
Notes
Incompatible with oxidizing agents.
Chemical Identifiers
998-93-6 | |
179.10 | |
BNUTXEKPXPZAIT-UHFFFAOYSA-N | |
70460 | |
CCCC(Br)CCC |
C7H15Br | |
MFCD00039182 | |
heptane, 4-bromo, 4-bromo-heptane, acmc-20ammr, 4-bromoheptane 5g | |
4-bromoheptane |
Specifications
998-93-6 | |
60°C to 62°C (18 mmHg) | |
C7H15Br | |
MFCD00039182 | |
UN1993 | |
heptane, 4-bromo, 4-bromo-heptane, acmc-20ammr, 4-bromoheptane 5g | |
BNUTXEKPXPZAIT-UHFFFAOYSA-N | |
4-bromoheptane | |
70460 | |
97% |
1.14 | |
40°C (104°F) | |
1.45 | |
5 g | |
1697144 | |
Miscible with carbon tetrachloride,chloroform and benzene. Immiscible with water. | |
CCCC(Br)CCC | |
179.10 | |
179.11 | |
4-Bromoheptane |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 213-653-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only