missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromoisophthalic acid, 96%, Thermo Scientific Chemicals

$129.29 - $1619.82

Chemical Identifiers

CAS 6939-93-1
Molecular Formula C8H5BrO4
Molecular Weight (g/mol) 245.028
MDL Number MFCD00002404
InChI Key MSQIEZXCNYUWHN-UHFFFAOYSA-N
Synonym 4-bromoisophthalic acid, 1,3-benzenedicarboxylic acid, 4-bromo, 4-bromoisophthalicacid, 4-bromo-1,3-benzenedicarboxylic acid, 2-bromobenzene-1,5-dicarboxylic acid, pubchem12479, chemwish ic03530, ksc354c4d, msqiezxcnyuwhn-uhfffaoysa
PubChem CID 81361
IUPAC Name 4-bromobenzene-1,3-dicarboxylic acid
SMILES C1=CC(=C(C=C1C(=O)O)C(=O)O)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1315206
View Documents
Thermo Scientific Chemicals
A1315206
5 g
Each for $129.29
 
AAA1315214
View Documents
Thermo Scientific Chemicals
A1315214
25 g
Each for $493.07
 
AAA1315222
View Documents
Thermo Scientific Chemicals
A1315222
100 g
Each for $1,619.82
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6939-93-1
245.028
MSQIEZXCNYUWHN-UHFFFAOYSA-N
81361
C1=CC(=C(C=C1C(=O)O)C(=O)O)Br
C8H5BrO4
MFCD00002404
4-bromoisophthalic acid, 1,3-benzenedicarboxylic acid, 4-bromo, 4-bromoisophthalicacid, 4-bromo-1,3-benzenedicarboxylic acid, 2-bromobenzene-1,5-dicarboxylic acid, pubchem12479, chemwish ic03530, ksc354c4d, msqiezxcnyuwhn-uhfffaoysa
4-bromobenzene-1,3-dicarboxylic acid

Specifications

6939-93-1
C8H5BrO4
5 g
4-bromoisophthalic acid, 1,3-benzenedicarboxylic acid, 4-bromo, 4-bromoisophthalicacid, 4-bromo-1,3-benzenedicarboxylic acid, 2-bromobenzene-1,5-dicarboxylic acid, pubchem12479, chemwish ic03530, ksc354c4d, msqiezxcnyuwhn-uhfffaoysa
C1=CC(=C(C=C1C(=O)O)C(=O)O)Br
245.028
245.03
4-Bromoisophthalic acid
296°C to 302°C
MFCD00002404
1959392
MSQIEZXCNYUWHN-UHFFFAOYSA-N
4-bromobenzene-1,3-dicarboxylic acid
81361
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 230-078-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only