missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
$202.71 - $699.42
Chemical Identifiers
CAS | 1029439-49-3 |
---|---|
Molecular Formula | C13H17BBrFO2 |
Molecular Weight (g/mol) | 314.989 |
MDL Number | MFCD18434471 |
InChI Key | QOCYLABQSVWXGE-UHFFFAOYSA-N |
Synonym | 2-4-bromomethyl-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-bromomethyl-2-fluorophenylboronic acid pinacol ester, 4-bromomethyl-2-fluorobenzeneboronic acid pinacol ester, 4-bromomethyl-2-fluorophenyl boronic acid pinacol ester |
PubChem CID | 53217331 |
IUPAC Name | 2-[4-(bromomethyl)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)CBr)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH59821MD
|
Thermo Scientific Chemicals
H59821MD |
250 mg |
Each for $202.71
|
|
||||||
AAH5982103
|
Thermo Scientific Chemicals
H5982103 |
1 g |
Each for $699.42
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1029439-49-3 | |
314.989 | |
QOCYLABQSVWXGE-UHFFFAOYSA-N | |
53217331 | |
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)CBr)F |
C13H17BBrFO2 | |
MFCD18434471 | |
2-4-bromomethyl-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-bromomethyl-2-fluorophenylboronic acid pinacol ester, 4-bromomethyl-2-fluorobenzeneboronic acid pinacol ester, 4-bromomethyl-2-fluorophenyl boronic acid pinacol ester | |
2-[4-(bromomethyl)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Specifications
1029439-49-3 | |
C13H17BBrFO2 | |
250 mg | |
QOCYLABQSVWXGE-UHFFFAOYSA-N | |
2-[4-(bromomethyl)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
53217331 | |
96% |
62°C to 64°C | |
MFCD18434471 | |
2-4-bromomethyl-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-bromomethyl-2-fluorophenylboronic acid pinacol ester, 4-bromomethyl-2-fluorobenzeneboronic acid pinacol ester, 4-bromomethyl-2-fluorophenyl boronic acid pinacol ester | |
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)CBr)F | |
314.989 | |
314.99 | |
4-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only