missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Bromomethyl)-3-methyl-5-phenylisoxazole, ≥95%, Thermo Scientific™

$456.24 - $456.24

Chemical Identifiers

CAS 113841-59-1
Molecular Formula C11H10BrNO
Molecular Weight (g/mol) 252.111
MDL Number MFCD03086124
InChI Key KCGVZUOBUPWFJC-UHFFFAOYSA-N
PubChem CID 2776518
IUPAC Name 4-(bromomethyl)-3-methyl-5-phenyl-1,2-oxazole
SMILES CC1=NOC(=C1CBr)C2=CC=CC=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC20308DA
SDS View Documents
Thermo Scientific Chemicals
CC20308DA
1 g Amber Glass Bottle
Each for $456.24
 
Description

Description

Specifications

Chemical Identifiers

113841-59-1
252.111
KCGVZUOBUPWFJC-UHFFFAOYSA-N
4-(bromomethyl)-3-methyl-5-phenyl-1,2-oxazole
C11H10BrNO
MFCD03086124
2776518
CC1=NOC(=C1CBr)C2=CC=CC=C2

Specifications

113841-59-1
100.0
Amber Glass Bottle
MFCD03086124
4-bromomethyl-3-methyl-5-phenylisoxazole, 4-bromomethyl-3-methyl-5-phenyl-1,2-oxazole, pubchem23336, acmc-1c8sr, 4-bromomethyl-3-methyl-5-phenyl-isoxazole, isoxazole,4-bromomethyl-3-methyl-5-phenyl, isoxazole, 4-bromomethyl-3-methyl-5-phenyl
CC1=NOC(=C1CBr)C2=CC=CC=C2
252.111
252.11
4-(Bromomethyl)-3-methyl-5-phenylisoxazole
94.5
95%
C11H10BrNO
1 g
KCGVZUOBUPWFJC-UHFFFAOYSA-N
4-(bromomethyl)-3-methyl-5-phenyl-1,2-oxazole
2776518
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'