Learn More
4-Bromomethyl-3-nitrobenzoic acid, 97%, Thermo Scientific Chemicals
$77.78 - $911.26
Chemical Identifiers
CAS | 55715-03-2 |
---|---|
Molecular Formula | C8H6BrNO4 |
Molecular Weight (g/mol) | 260.043 |
MDL Number | MFCD00216575 |
InChI Key | QMAHVAFURJBOFV-UHFFFAOYSA-N |
Synonym | 4-bromomethyl-3-nitrobenzoic acid, 4-bromomethyl-3-nitrobenzoicacid, benzoic acid, 4-bromomethyl-3-nitro, 3-nitro-4-bromomethylbenzoic acid, bnba, acmc-1auor, ksc495i9f, 3-nitro-4-bromomethyl benzoic acid, 4-bromomethyl-3-nitro benzoic acid |
PubChem CID | 132386 |
SMILES | C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])CBr |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAB2525503
|
Thermo Scientific Chemicals
B2525503 |
1 g |
Each for $77.78
|
|
||||||
AAB2525506
|
Thermo Scientific Chemicals
B2525506 |
5 g |
Each for $250.19
|
|
||||||
AAB2525514
|
Thermo Scientific Chemicals
B2525514 |
25 g |
Each for $911.26
|
|
||||||
Description
4-Bromomethyl-3-nitrobenzoic acid is used as a reactant in the synthesis of 4-bromomethyl-3-nitrobenzoic acid succinimide ester, 4-((2-(hydroxymethyl)phenyl amino)methyl)-3-nitrobenzoic acid, 4-(2-hydroxyethylmercaptylmethyl)-3-nitrobenzoic acid. It is also used as a thiol photo-deprotection reagent, a starting material in the synthesis of 2H-indazole based library using parallel solution-phase methods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromomethyl-3-nitrobenzoic acid is used as a reactant in the synthesis of 4-bromomethyl-3-nitrobenzoic acid succinimide ester, 4-((2-(hydroxymethyl)phenyl amino)methyl)-3-nitrobenzoic acid, 4-(2-hydroxyethylmercaptylmethyl)-3-nitrobenzoic acid. It is also used as a thiol photo-deprotection reagent, a starting material in the synthesis of 2H-indazole based library using parallel solution-phase methods.
Solubility
Soluble in DMF and dichloromethane. Insoluble in water.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents, bases.
Chemical Identifiers
55715-03-2 | |
260.043 | |
QMAHVAFURJBOFV-UHFFFAOYSA-N | |
132386 |
C8H6BrNO4 | |
MFCD00216575 | |
4-bromomethyl-3-nitrobenzoic acid, 4-bromomethyl-3-nitrobenzoicacid, benzoic acid, 4-bromomethyl-3-nitro, 3-nitro-4-bromomethylbenzoic acid, bnba, acmc-1auor, ksc495i9f, 3-nitro-4-bromomethyl benzoic acid, 4-bromomethyl-3-nitro benzoic acid | |
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])CBr |
Specifications
55715-03-2 | |
C8H6BrNO4 | |
1 g | |
4-bromomethyl-3-nitrobenzoic acid, 4-bromomethyl-3-nitrobenzoicacid, benzoic acid, 4-bromomethyl-3-nitro, 3-nitro-4-bromomethylbenzoic acid, bnba, acmc-1auor, ksc495i9f, 3-nitro-4-bromomethyl benzoic acid, 4-bromomethyl-3-nitro benzoic acid | |
QMAHVAFURJBOFV-UHFFFAOYSA-N | |
260.043 | |
260.05 | |
4-Bromomethyl-3-nitrobenzoic acid |
128°C to 133°C | |
MFCD00216575 | |
UN3261 | |
Soluble in DMF and dichloromethane. Insoluble in water. | |
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])CBr | |
132386 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only