missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Bromomethyl)-5-methyl-3-phenylisoxazole, 97%, Thermo Scientific™

$201.47 - $281.92

Chemical Identifiers

CAS 180597-83-5
Molecular Formula C11H10BrNO
Molecular Weight (g/mol) 252.11
MDL Number MFCD02677680
InChI Key UICMWXWMCOJBIQ-UHFFFAOYSA-N
PubChem CID 2776147
IUPAC Name 4-(bromomethyl)-5-methyl-3-phenyl-1,2-oxazole
SMILES CC1=C(CBr)C(=NO1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC00508CB
SDS View Documents
Thermo Scientific Chemicals
CC00508CB
250 mg N/A
 
CC00508DA
SDS View Documents
Thermo Scientific Chemicals
CC00508DA
1 g N/A
 
Description

Description

Specifications

Chemical Identifiers

180597-83-5
252.11
UICMWXWMCOJBIQ-UHFFFAOYSA-N
4-(bromomethyl)-5-methyl-3-phenyl-1,2-oxazole
C11H10BrNO
MFCD02677680
2776147
CC1=C(CBr)C(=NO1)C1=CC=CC=C1

Specifications

180597-83-5
100.0
Amber Glass Bottle
MFCD02677680
4-bromomethyl-5-methyl-3-phenylisoxazole, 4-bromomethyl-5-methyl-3-phenyl-1,2-oxazole, isoxazole, 4-bromomethyl-5-methyl-3-phenyl, 4-bromomethyl-5-methyl-3-phenylisoxazol, isoxazole,4-bromomethyl-5-methyl-3-phenyl
CC1=C(CBr)C(=NO1)C1=CC=CC=C1
252.11
252.11
4-(Bromomethyl)-5-methyl-3-phenylisoxazole
96.5
97%
C11H10BrNO
250 mg
UICMWXWMCOJBIQ-UHFFFAOYSA-N
4-(bromomethyl)-5-methyl-3-phenyl-1,2-oxazole
2776147
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'