missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(Bromomethyl)benzoic Acid 97.0+%, TCI America™
Supplier: TCI America B084525G
Specifications
4-(Bromomethyl)benzoic Acid | |
230°C | |
C8H7BrO2 | |
25 g | |
4-bromomethyl benzoic acid, alpha-bromo-p-toluic acid, benzoic acid, 4-bromomethyl, p-bromomethyl benzoic acid, 4-carboxybenzyl bromide, 4-bromomethyl-benzoic acid, a-bromo-p-toluic acid, p-carboxybenzyl bromide, p-bromomethylbenzoic acid | |
C1=CC(=CC=C1CBr)C(=O)O | |
215.046 | |
215.05 | |
Crystalline Powder |
6232-88-8 | |
White | |
MFCD00002567 | |
3261 | |
CQQSQBRPAJSTFB-UHFFFAOYSA-N | |
4-(bromomethyl)benzoic acid | |
22599 | |
≥97.0% (GC,T) |
Chemical Identifiers
6232-88-8 | |
215.046 | |
CQQSQBRPAJSTFB-UHFFFAOYSA-N | |
22599 | |
C1=CC(=CC=C1CBr)C(=O)O |
C8H7BrO2 | |
MFCD00002567 | |
4-bromomethyl benzoic acid, alpha-bromo-p-toluic acid, benzoic acid, 4-bromomethyl, p-bromomethyl benzoic acid, 4-carboxybenzyl bromide, 4-bromomethyl-benzoic acid, a-bromo-p-toluic acid, p-carboxybenzyl bromide, p-bromomethylbenzoic acid | |
4-(bromomethyl)benzoic acid |
Safety and Handling
TSCA : Yes