missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Bromomethyl)phenylacetic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2774406

 View more versions of this product

Catalog No. AAH2774406


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-(Bromomethyl)phenylacetic acid
178°C to 180°C
MFCD00010632
UN3261
WCOCCXZFEJGHTC-UHFFFAOYSA-N
2-[4-(bromomethyl)phenyl]acetic acid
4519056
97%
13737-36-5
C9H9BrO2
5 g
4-bromomethyl phenylacetic acid, 4-bromomethylphenylacetic acid, 2-4-bromomethyl phenyl acetic acid, pam acid, br-pam-linker, 4-carboxymethyl benzyl bromide, 4-bromomethyl phenyl acetic acid, 4-bromomethyl phenylaceticacid
C1=CC(=CC=C1CC(=O)O)CBr
229.073
229.07

Chemical Identifiers

13737-36-5
229.073
WCOCCXZFEJGHTC-UHFFFAOYSA-N
4519056
C1=CC(=CC=C1CC(=O)O)CBr
C9H9BrO2
MFCD00010632
4-bromomethyl phenylacetic acid, 4-bromomethylphenylacetic acid, 2-4-bromomethyl phenyl acetic acid, pam acid, br-pam-linker, 4-carboxymethyl benzyl bromide, 4-bromomethyl phenyl acetic acid, 4-bromomethyl phenylaceticacid
2-[4-(bromomethyl)phenyl]acetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H332-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only