missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
4-(Bromomethyl)tetrahydropyran, 97%, Thermo Scientific™
Chemical Identifiers
CAS |
125552-89-8 |
Molecular Formula |
C6H11BrO |
Molecular Weight (g/mol) |
179.057 |
InChI Key |
LMOOYAKLEOGKJR-UHFFFAOYSA-N |
Synonym |
4-bromomethyl tetrahydropyran, 4-bromomethyltetrahydropyran, 4-bromomethyl oxane, 4-bromomethyl tetrahydro-2h-pyran, 4-bromomethyl-tetrahydro-pyran, 4-bromomethyl tetrahydro-pyran, 4-bromomethyl-tetrahydro-2h-pyran, 4-bromomethyl-tetrahydropyran, 2h-pyran, 4-bromomethyl tetrahydro |
PubChem CID |
2773286 |
IUPAC Name |
4-(bromomethyl)oxane |
SMILES |
C1COCCC1CBr |
View More Specs
Products
1
Catalog Number |
Mfr. No. |
Quantity |
Packaging |
Price |
Quantity |
|
|
|
|
CC29908DA
SDS
View Documents
|
Thermo Scientific Chemicals
CC29908DA
|
1 g |
Amber Glass Bottle |
|
|
Add to cart
|
|
|
|
Description
Specifications
Chemical Identifiers
|
125552-89-8 |
|
179.057 |
|
4-bromomethyl tetrahydropyran, 4-bromomethyltetrahydropyran, 4-bromomethyl oxane, 4-bromomethyl tetrahydro-2h-pyran, 4-bromomethyl-tetrahydro-pyran, 4-bromomethyl tetrahydro-pyran, 4-bromomethyl-tetrahydro-2h-pyran, 4-bromomethyl-tetrahydropyran, 2h-pyran, 4-bromomethyl tetrahydro |
|
4-(bromomethyl)oxane |
|
C6H11BrO |
|
LMOOYAKLEOGKJR-UHFFFAOYSA-N |
|
2773286 |
|
C1COCCC1CBr |
Specifications
|
125552-89-8 |
|
Amber Glass Bottle |
|
1 g |
|
LMOOYAKLEOGKJR-UHFFFAOYSA-N |
|
4-(bromomethyl)oxane |
|
2773286 |
|
97% |
|
97% |
|
C6H11BrO |
|
4-bromomethyl tetrahydropyran, 4-bromomethyltetrahydropyran, 4-bromomethyl oxane, 4-bromomethyl tetrahydro-2h-pyran, 4-bromomethyl-tetrahydro-pyran, 4-bromomethyl tetrahydro-pyran, 4-bromomethyl-tetrahydro-2h-pyran, 4-bromomethyl-tetrahydropyran, 2h-pyran, 4-bromomethyl tetrahydro |
|
C1COCCC1CBr |
|
179.057 |
|
179.06 |
|
4-(Bromomethyl)tetrahydropyran |
Videos
SDS
missing translation for 'documents'