missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromophenol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1487336

 View more versions of this product

Catalog No. AAA1487336


Explore available promotions

Description

Description

4-Bromophenol is used as an intermediate in medicine grade. It is also used as a phenol that is used in Stille reactions. It is used in the intermediate of organic synthesis, flame retardant and pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromophenol is used as an intermediate in medicine grade. It is also used as a phenol that is used in Stille reactions. It is used in the intermediate of organic synthesis, flame retardant and pharmaceutical industry.

Solubility
It is insoluble in water. It is soluble in 5%ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Store under inert gas. Air sensitive. Keep away from Oxidizing agents.
Specifications

Specifications

4-Bromophenol
62°C to 66°C
235°C to 238°C
99%
C6H5BrO
500 g
1680024
14,1428
It is insoluble in water. It is soluble in 5%ethanol.
C1=CC(=CC=C1O)Br
173.009
173.01
106-41-2
1.84
>110°C (230°F)
Phenol-like
MFCD00002313
UN2811
Light sensitive
p-bromophenol, phenol, 4-bromo, p-bromohydroxybenzene, 4-bromo-phenol, p-bromophenic acid, phenol, p-bromo, para-bromophenol, 4-bromo phenol, unii-lao4j0183i, ccris 632
GZFGOTFRPZRKDS-UHFFFAOYSA-N
4-bromophenol
7808
99%

Chemical Identifiers

106-41-2
173.009
GZFGOTFRPZRKDS-UHFFFAOYSA-N
7808
C1=CC(=CC=C1O)Br
C6H5BrO
MFCD00002313
p-bromophenol, phenol, 4-bromo, p-bromohydroxybenzene, 4-bromo-phenol, p-bromophenic acid, phenol, p-bromo, para-bromophenol, 4-bromo phenol, unii-lao4j0183i, ccris 632
4-bromophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 203-394-4

RTECSNumber : SJ7960000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only