missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Bromophenoxyacetic acid, 95%, Thermo Scientific™
$213.66 - $213.66
Chemical Identifiers
CAS | 1878-91-7 |
---|---|
Molecular Formula | C8H7BrO3 |
Molecular Weight (g/mol) | 231.045 |
MDL Number | MFCD00093070 |
InChI Key | SZEBGAQWWSUOHT-UHFFFAOYSA-N |
Synonym | 4-bromophenoxyacetic acid, 2-4-bromophenoxy acetic acid, 4-bromophenoxy acetic acid, p-bromophenoxyacetic acid, acetic acid, 4-bromophenoxy, p-bromophenoxyacetate, p-bromophenoxyacetic acvid, acetic acid, 2-4-bromophenoxy, acetic acid, p-bromophenoxy |
PubChem CID | 74657 |
IUPAC Name | 2-(4-bromophenoxy)acetic acid |
SMILES | C1=CC(=CC=C1OCC(=O)O)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6088814
|
Thermo Scientific Chemicals
H6088814 |
25 g |
Each for $213.66
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1878-91-7 | |
231.045 | |
SZEBGAQWWSUOHT-UHFFFAOYSA-N | |
74657 | |
C1=CC(=CC=C1OCC(=O)O)Br |
C8H7BrO3 | |
MFCD00093070 | |
4-bromophenoxyacetic acid, 2-4-bromophenoxy acetic acid, 4-bromophenoxy acetic acid, p-bromophenoxyacetic acid, acetic acid, 4-bromophenoxy, p-bromophenoxyacetate, p-bromophenoxyacetic acvid, acetic acid, 2-4-bromophenoxy, acetic acid, p-bromophenoxy | |
2-(4-bromophenoxy)acetic acid |
Specifications
1878-91-7 | |
C8H7BrO3 | |
25 g | |
SZEBGAQWWSUOHT-UHFFFAOYSA-N | |
2-(4-bromophenoxy)acetic acid | |
74657 | |
95% |
149°C to 153°C | |
MFCD00093070 | |
4-bromophenoxyacetic acid, 2-4-bromophenoxy acetic acid, 4-bromophenoxy acetic acid, p-bromophenoxyacetic acid, acetic acid, 4-bromophenoxy, p-bromophenoxyacetate, p-bromophenoxyacetic acvid, acetic acid, 2-4-bromophenoxy, acetic acid, p-bromophenoxy | |
C1=CC(=CC=C1OCC(=O)O)Br | |
231.045 | |
231.04 | |
4-Bromophenoxyacetic acid |
Safety and Handling
EINECSNumber : 217-530-5
TSCA : Yes
Recommended Storage : Ambient temperatures