Learn More
4-Bromophenylacetonitrile, 99%, Thermo Scientific Chemicals
$54.47 - $54.47
Chemical Identifiers
CAS | 16532-79-9 |
---|---|
Molecular Formula | C8H6BrN |
Molecular Weight (g/mol) | 196.05 |
MDL Number | MFCD00001916 |
InChI Key | MFHFWRBXPQDZSA-UHFFFAOYSA-N |
Synonym | 4-bromophenylacetonitrile, 4-bromobenzyl cyanide, 2-4-bromophenyl acetonitrile, benzeneacetonitrile, 4-bromo, p-bromophenylacetonitrile, 4-bromobenzylcyanide, 4-bromobenzeneacetonitrile, p-bromobenzyl cyanide, p-bromobenzylcyanide, acetonitrile, p-bromophenyl |
PubChem CID | 27914 |
IUPAC Name | 2-(4-bromophenyl)acetonitrile |
SMILES | BrC1=CC=C(CC#N)C=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC154480250
|
Thermo Scientific Chemicals
154480250 |
25 g | Glass bottle |
Each for $54.47
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16532-79-9 | |
196.05 | |
MFHFWRBXPQDZSA-UHFFFAOYSA-N | |
27914 | |
BrC1=CC=C(CC#N)C=C1 |
C8H6BrN | |
MFCD00001916 | |
4-bromophenylacetonitrile, 4-bromobenzyl cyanide, 2-4-bromophenyl acetonitrile, benzeneacetonitrile, 4-bromo, p-bromophenylacetonitrile, 4-bromobenzylcyanide, 4-bromobenzeneacetonitrile, p-bromobenzyl cyanide, p-bromobenzylcyanide, acetonitrile, p-bromophenyl | |
2-(4-bromophenyl)acetonitrile |
Specifications
16532-79-9 | |
100.0 | |
Brown to Colorless | |
Authentic | |
Glass bottle | |
BrC6H4CH2CN | |
25 g | |
4-bromophenylacetonitrile, 4-bromobenzyl cyanide, 2-4-bromophenyl acetonitrile, benzeneacetonitrile, 4-bromo, p-bromophenylacetonitrile, 4-bromobenzylcyanide, 4-bromobenzeneacetonitrile, p-bromobenzyl cyanide, p-bromobenzylcyanide, acetonitrile, p-bromophenyl | |
MFHFWRBXPQDZSA-UHFFFAOYSA-N | |
2-(4-bromophenyl)acetonitrile | |
27914 | |
99% | |
4-Bromophenylacetonitrile, 99% |
98.5 | |
47°C to 49°C | |
152°C | |
98.5% min. (GC) | |
C8H6BrN | |
MFCD00001916 | |
09, 451 | |
Solubility in water: insoluble | |
BrC1=CC=C(CC#N)C=C1 | |
196.05 | |
196.05 | |
Crystalline Mass |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Toxic if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If eye irr
GHS Signal Word: Danger
EINECSNumber : 240-602-2
RTECSNumber : AL8090000
TSCA : TSCA
RUO – Research Use Only