missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(4-Bromophenylethynyl)trimethylsilane, 98%, Thermo Scientific Chemicals
$55.37 - $682.86
Chemical Identifiers
CAS | 16116-78-2 |
---|---|
Molecular Formula | C11H13BrSi |
Molecular Weight (g/mol) | 253.21 |
MDL Number | MFCD01863556 |
InChI Key | RNMSGCJGNJYDNS-UHFFFAOYSA-N |
Synonym | 4-bromophenylethynyl trimethylsilane, 4-bromophenyl ethynyl trimethylsilane, 4-bromo-phenylethynyl-trimethyl-silane, 2-4-bromophenyl ethynyl trimethylsilane, acmc-1bvav, p-bromophenylethynyl trimethylsilane, 4-trimethylsilylethynyl bromo benzene, 4-bromo-phenylethynyl trimethyl silane |
PubChem CID | 4226980 |
IUPAC Name | 2-(4-bromophenyl)ethynyl-trimethylsilane |
SMILES | C[Si](C)(C)C#CC1=CC=C(Br)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5553403
|
Thermo Scientific Chemicals
H5553403 |
1 g |
Each for $55.37
|
|
|||||
AAH5553406
|
Thermo Scientific Chemicals
H5553406 |
5 g |
Each for $185.03
|
|
|||||
AAH5553414
|
Thermo Scientific Chemicals
H5553414 |
25 g |
Each for $682.86
|
|
|||||
Description
It is used as the intermediate of Organic Light Emitting Diode (OLED).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as the intermediate of Organic Light Emitting Diode (OLED).
Notes
Store at room temperature. Avoid strong oxidizing agents.
Chemical Identifiers
16116-78-2 | |
253.21 | |
RNMSGCJGNJYDNS-UHFFFAOYSA-N | |
4226980 | |
C[Si](C)(C)C#CC1=CC=C(Br)C=C1 |
C11H13BrSi | |
MFCD01863556 | |
4-bromophenylethynyl trimethylsilane, 4-bromophenyl ethynyl trimethylsilane, 4-bromo-phenylethynyl-trimethyl-silane, 2-4-bromophenyl ethynyl trimethylsilane, acmc-1bvav, p-bromophenylethynyl trimethylsilane, 4-trimethylsilylethynyl bromo benzene, 4-bromo-phenylethynyl trimethyl silane | |
2-(4-bromophenyl)ethynyl-trimethylsilane |
Specifications
61°C to 63°C | |
16116-78-2 | |
1 g | |
MFCD01863556 | |
RNMSGCJGNJYDNS-UHFFFAOYSA-N | |
2-(4-bromophenyl)ethynyl-trimethylsilane | |
4226980 | |
98% |
White to Yellow | |
Powder | |
C11H13BrSi | |
4-bromophenylethynyl trimethylsilane, 4-bromophenyl ethynyl trimethylsilane, 4-bromo-phenylethynyl-trimethyl-silane, 2-4-bromophenyl ethynyl trimethylsilane, acmc-1bvav, p-bromophenylethynyl trimethylsilane, 4-trimethylsilylethynyl bromo benzene, 4-bromo-phenylethynyl trimethyl silane | |
C[Si](C)(C)C#CC1=CC=C(Br)C=C1 | |
253.21 | |
253.21 | |
(4-Bromophenylethynyl)trimethylsilane |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only