missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromothioanisole, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1550436

 View more versions of this product

Catalog No. AAA1550436


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromothioanisole
37°C to 40°C
>110°C (230°F)
C7H7BrS
500 g
4-bromothioanisole, 1-bromo-4-methylthio benzene, p-bromophenyl methyl sulfide, 4-bromophenyl methyl sulfide, benzene, 1-bromo-4-methylthio, 1-bromo-4-methylsulfanyl benzene, p-bromothioanisole, 4-bromo-thioanisole, methyl 4-bromophenylsulfide, p-bromo thioanisole
CSC1=CC=C(Br)C=C1
203.10
203.11
104-95-0
250°C
Stench
MFCD00000102
1906693
YEUYZNNBXLMFCW-UHFFFAOYSA-N
1-bromo-4-methylsulfanylbenzene
66037
≥98%

Chemical Identifiers

104-95-0
203.10
YEUYZNNBXLMFCW-UHFFFAOYSA-N
66037
CSC1=CC=C(Br)C=C1
C7H7BrS
MFCD00000102
4-bromothioanisole, 1-bromo-4-methylthio benzene, p-bromophenyl methyl sulfide, 4-bromophenyl methyl sulfide, benzene, 1-bromo-4-methylthio, 1-bromo-4-methylsulfanyl benzene, p-bromothioanisole, 4-bromo-thioanisole, methyl 4-bromophenylsulfide, p-bromo thioanisole
1-bromo-4-methylsulfanylbenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Hazard Class: 6; Packaging Group: III

EINECSNumber : 203-255-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only