missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromothiobenzamide, 97%, Thermo Scientific Chemicals

$43.82 - $131.66

Chemical Identifiers

CAS 26197-93-3
Molecular Formula C7H6BrNS
Molecular Weight (g/mol) 216.096
MDL Number MFCD04114360
InChI Key AIPANIYQEBQYGC-UHFFFAOYSA-N
Synonym 4-bromothiobenzamide, 4-bromo-thiobenzamide, p-bromothiobenzamide, 4-bromobenzothioamide, benzenecarbothioamide, 4-bromo, 4-bromobenzene-1-carbothioamide, amino 4-bromophenyl methane-1-thione, acmc-1cehr, 4-bromanylbenzenecarbothioamide
PubChem CID 1549289
IUPAC Name 4-bromobenzenecarbothioamide
SMILES C1=CC(=CC=C1C(=S)N)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5595603
View Documents
Thermo Scientific Chemicals
H5595603
1 g
Each for $43.82
 
AAH5595606
View Documents
Thermo Scientific Chemicals
H5595606
5 g
Each for $131.66
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

26197-93-3
216.096
AIPANIYQEBQYGC-UHFFFAOYSA-N
1549289
C1=CC(=CC=C1C(=S)N)Br
C7H6BrNS
MFCD04114360
4-bromothiobenzamide, 4-bromo-thiobenzamide, p-bromothiobenzamide, 4-bromobenzothioamide, benzenecarbothioamide, 4-bromo, 4-bromobenzene-1-carbothioamide, amino 4-bromophenyl methane-1-thione, acmc-1cehr, 4-bromanylbenzenecarbothioamide
4-bromobenzenecarbothioamide

Specifications

26197-93-3
Brown to White
MFCD04114360
4-bromothiobenzamide, 4-bromo-thiobenzamide, p-bromothiobenzamide, 4-bromobenzothioamide, benzenecarbothioamide, 4-bromo, 4-bromobenzene-1-carbothioamide, amino 4-bromophenyl methane-1-thione, acmc-1cehr, 4-bromanylbenzenecarbothioamide
C1=CC(=CC=C1C(=S)N)Br
216.096
216.1
Solid
139°C to 143°C
C7H6BrNS
1 g
AIPANIYQEBQYGC-UHFFFAOYSA-N
4-bromobenzenecarbothioamide
1549289
97%
4-Bromothiobenzamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.

P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c

H302-H317

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only