missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Carboxymethyl)phenylboronic acid pinacol ester, 97%, Thermo Scientific™

Chemical Identifiers

CAS 797755-07-8
Molecular Formula C14H19BO4
Molecular Weight (g/mol) 262.11
MDL Number MFCD04038755
InChI Key FNLWHBHWDXCWHV-UHFFFAOYSA-N
Synonym 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetic acid, 4-carboxymethyl phenylboronic acid pinacol ester, phenylacetic acid-4-boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetic acid, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylacetate, benzeneacetic acid, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-dihydroxyborane phenylacetic acid pinacol ester, 4-carboxymethyl phenylboronic acidpinacol ester
PubChem CID 3719002
IUPAC Name 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(CC(O)=O)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC444670010
SDS View Documents
Thermo Scientific Chemicals
444670010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

797755-07-8
262.11
FNLWHBHWDXCWHV-UHFFFAOYSA-N
3719002
CC1(C)OB(OC1(C)C)C1=CC=C(CC(O)=O)C=C1
C14H19BO4
MFCD04038755
2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetic acid, 4-carboxymethyl phenylboronic acid pinacol ester, phenylacetic acid-4-boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetic acid, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylacetate, benzeneacetic acid, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-dihydroxyborane phenylacetic acid pinacol ester, 4-carboxymethyl phenylboronic acidpinacol ester
2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid

Specifications

797755-07-8
Glass Bottle
MFCD04038755
2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetic acid, 4-carboxymethyl phenylboronic acid pinacol ester, phenylacetic acid-4-boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetic acid, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylacetate, benzeneacetic acid, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-dihydroxyborane phenylacetic acid pinacol ester, 4-carboxymethyl phenylboronic acidpinacol ester
CC1(C)OB(OC1(C)C)C1=CC=C(CC(O)=O)C=C1
262.11
262.11
4-(Carboxymethyl)phenylboronic acid pinacol ester
97%
C14H19BO4
1g
FNLWHBHWDXCWHV-UHFFFAOYSA-N
2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid
3719002
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only